Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 49328509-49328734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM227B
Entrez Gene ID
196951
Gene Name
family with sequence similarity 227 member B
Gene Aliases
C15orf33
UniGene
Hs.712959
Gene Chromosome Location
Chr.15: 49326997-49620931 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521322.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 NM_001001556.2 NM_001289030.1 NM_001289031.1 NM_002044.3 XM_005254279.3 XM_005254280.3 XM_005254284.3 XM_006720461.3 XM_006720462.3 XM_006720463.3 XM_006720464.3 XM_011521441.2 XM_017022062.1 XM_017022063.1 XM_017022064.1 XM_017022065.1 XM_017022066.1

Gene Symbol
GALK2
Entrez Gene ID
2585
Gene Name
galactokinase 2
Gene Aliases
GK2
UniGene
Hs.122006
Gene Chromosome Location
Chr.15: 49155757-49367883 on Build GRCh38
Transcripts
NM_152647.2 XM_005254213.3 XM_005254214.3 XM_005254215.3 XM_005254216.3 XM_005254219.3 XM_006720423.2 XM_006720424.2 XM_006720426.2 XM_006720427.3 XM_011521319.2 XM_011521320.1 XM_011521322.1 XM_017021990.1 XM_017021991.1 XM_017021992.1 XM_017021993.1 XM_017021994.1 XM_017021995.1 XM_017021996.1 NM_001001556.2 NM_001289030.1 NM_001289031.1 NM_002044.3 XM_005254279.3 XM_005254280.3 XM_005254284.3 XM_006720461.3 XM_006720462.3 XM_006720463.3 XM_006720464.3 XM_011521441.2 XM_017022062.1 XM_017022063.1 XM_017022064.1 XM_017022065.1 XM_017022066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
433997 c.1456T>A p.S486T Chr.15: 49328639-49328639 on Build GRCh38
5768591 c.1522T>G p.Y508D Chr.15: 49328573-49328573 on Build GRCh38
1176705 c.1426G>A p.E476K Chr.15: 49328669-49328669 on Build GRCh38
7447933 c.1483_1491delCCATCATCA p.P495_S497delPSS Chr.15: 49328604-49328612 on Build GRCh38
7204412 c.1465_1467delTCA p.S494delS Chr.15: 49328628-49328630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150753064 Chr.15: 49328671-49328671 on Build GRCh38
rs761403515 Chr.15: 49328696-49328696 on Build GRCh38
rs774318678 Chr.15: 49328674-49328674 on Build GRCh38
rs773063173 Chr.15: 49328636-49328636 on Build GRCh38
rs772992443 Chr.15: 49328623-49328631 on Build GRCh38
rs753592849 Chr.15: 49328550-49328550 on Build GRCh38
rs747851240 Chr.15: 49328613-49328618 on Build GRCh38
rs769958202 Chr.15: 49328629-49328631 on Build GRCh38
rs758303417 Chr.15: 49328595-49328595 on Build GRCh38
rs758836221 Chr.15: 49328604-49328606 on Build GRCh38
rs746272410 Chr.15: 49328651-49328651 on Build GRCh38
rs750239325 Chr.15: 49328584-49328584 on Build GRCh38
rs927481555 Chr.15: 49328612-49328612 on Build GRCh38
rs776241001 Chr.15: 49328612-49328613 on Build GRCh38
rs754154694 Chr.15: 49328574-49328576 on Build GRCh38
rs779561225 Chr.15: 49328604-49328612 on Build GRCh38
rs749471661 Chr.15: 49328628-49328628 on Build GRCh38
rs141143208 Chr.15: 49328596-49328596 on Build GRCh38
rs761376123 Chr.15: 49328568-49328568 on Build GRCh38
rs367997810 Chr.15: 49328576-49328576 on Build GRCh38
rs768317870 Chr.15: 49328613-49328615 on Build GRCh38
rs757674601 Chr.15: 49328596-49328597 on Build GRCh38
rs756949145 Chr.15: 49328607-49328612 on Build GRCh38
rs746374746 Chr.15: 49328604-49328624 on Build GRCh38
rs752706367 Chr.15: 49328609-49328609 on Build GRCh38
rs766287938 Chr.15: 49328670-49328670 on Build GRCh38
rs776016532 Chr.15: 49328675-49328675 on Build GRCh38
rs771303406 Chr.15: 49328631-49328631 on Build GRCh38
rs184970229 Chr.15: 49328617-49328617 on Build GRCh38
rs77994602 Chr.15: 49328579-49328579 on Build GRCh38
rs368311811 Chr.15: 49328700-49328700 on Build GRCh38
rs762888521 Chr.15: 49328650-49328653 on Build GRCh38
rs779480779 Chr.15: 49328603-49328604 on Build GRCh38
rs375041430 Chr.15: 49328574-49328574 on Build GRCh38
rs373923791 Chr.15: 49328669-49328669 on Build GRCh38
rs960806833 Chr.15: 49328649-49328649 on Build GRCh38
rs769416028 Chr.15: 49328580-49328580 on Build GRCh38
rs958929073 Chr.15: 49328647-49328647 on Build GRCh38

View Full Product Details