Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17772404-17772665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCHO1
Entrez Gene ID
23149
Gene Name
FCH domain only 1
Gene Aliases
-
UniGene
Hs.96485
Gene Chromosome Location
Chr.19: 17747718-17788568 on Build GRCh38
Transcripts
NM_001161357.1 NM_001161358.1 NM_001161359.1 NM_015122.2 XM_005259831.3 XM_006722701.3 XM_006722702.3 XM_011527824.2 XM_011527825.2 XM_011527826.2 XM_011527827.2 XM_011527828.2 XM_017026519.1 XM_017026520.1 XM_017026521.1 XM_017026522.1 XM_017026523.1 XM_017026524.1 XM_017026525.1 XM_017026526.1 XM_017026527.1 XM_017026528.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
320390 c.712C>A p.Q238K Chr.19: 17772663-17772663 on Build GRCh38
6662778 c.679G>A p.V227M Chr.19: 17772541-17772541 on Build GRCh38
438931 c.601C>T p.Q201* Chr.19: 17772463-17772463 on Build GRCh38
5330166 c.615G>A p.E205E Chr.19: 17772477-17772477 on Build GRCh38
4075473 c.675G>T p.T225T Chr.19: 17772537-17772537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908538056 Chr.19: 17772432-17772432 on Build GRCh38
rs761515469 Chr.19: 17772448-17772448 on Build GRCh38
rs749569094 Chr.19: 17772540-17772540 on Build GRCh38
rs766269522 Chr.19: 17772480-17772480 on Build GRCh38
rs372042106 Chr.19: 17772525-17772525 on Build GRCh38
rs754460155 Chr.19: 17772487-17772487 on Build GRCh38
rs776899624 Chr.19: 17772430-17772430 on Build GRCh38
rs760418723 Chr.19: 17772565-17772565 on Build GRCh38
rs760655505 Chr.19: 17772475-17772475 on Build GRCh38
rs919450995 Chr.19: 17772551-17772551 on Build GRCh38
rs763808870 Chr.19: 17772629-17772629 on Build GRCh38
rs762747824 Chr.19: 17772628-17772628 on Build GRCh38
rs914599625 Chr.19: 17772481-17772481 on Build GRCh38
rs368303042 Chr.19: 17772440-17772440 on Build GRCh38
rs149164146 Chr.19: 17772630-17772630 on Build GRCh38
rs147764124 Chr.19: 17772552-17772552 on Build GRCh38
rs867511648 Chr.19: 17772457-17772457 on Build GRCh38
rs752330518 Chr.19: 17772489-17772489 on Build GRCh38
rs528053341 Chr.19: 17772536-17772536 on Build GRCh38
rs767365658 Chr.19: 17772458-17772458 on Build GRCh38
rs373999164 Chr.19: 17772569-17772569 on Build GRCh38
rs1036103313 Chr.19: 17772541-17772541 on Build GRCh38
rs781696574 Chr.19: 17772518-17772518 on Build GRCh38
rs772977846 Chr.19: 17772470-17772470 on Build GRCh38
rs977363948 Chr.19: 17772517-17772517 on Build GRCh38
rs764812867 Chr.19: 17772488-17772488 on Build GRCh38
rs144207911 Chr.19: 17772486-17772486 on Build GRCh38
rs371722255 Chr.19: 17772602-17772602 on Build GRCh38
rs779991590 Chr.19: 17772537-17772537 on Build GRCh38
rs758122344 Chr.19: 17772502-17772502 on Build GRCh38
rs28461486 Chr.19: 17772574-17772574 on Build GRCh38
rs758976400 Chr.19: 17772592-17772592 on Build GRCh38
rs749728950 Chr.19: 17772589-17772589 on Build GRCh38
rs188192228 Chr.19: 17772425-17772425 on Build GRCh38
rs755969650 Chr.19: 17772568-17772569 on Build GRCh38
rs757880771 Chr.19: 17772514-17772514 on Build GRCh38
rs778343598 Chr.19: 17772504-17772504 on Build GRCh38
rs775733974 Chr.19: 17772441-17772441 on Build GRCh38
rs768844335 Chr.19: 17772546-17772546 on Build GRCh38
rs751002998 Chr.19: 17772638-17772638 on Build GRCh38
rs752139684 Chr.19: 17772609-17772609 on Build GRCh38
rs570592690 Chr.19: 17772637-17772637 on Build GRCh38
rs376998937 Chr.19: 17772438-17772438 on Build GRCh38
rs531090742 Chr.19: 17772524-17772524 on Build GRCh38
rs751949561 Chr.19: 17772505-17772505 on Build GRCh38

View Full Product Details