Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 47090267-47090520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf61
Entrez Gene ID
285051
Gene Name
chromosome 2 open reading frame 61
Gene Aliases
-
UniGene
Hs.531575
Gene Chromosome Location
Chr.2: 47086991-47155378 on Build GRCh38
Transcripts
NM_001163561.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6311732 c.559C>A p.P187T Chr.2: 47090335-47090335 on Build GRCh38
1021228 c.614C>T p.P205L Chr.2: 47090280-47090280 on Build GRCh38
5058919 c.537A>G p.P179P Chr.2: 47090357-47090357 on Build GRCh38
4001690 c.520-3T>A p.? Chr.2: 47090377-47090377 on Build GRCh38
3185814 c.600C>T p.V200V Chr.2: 47090294-47090294 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs547416570 Chr.2: 47090460-47090460 on Build GRCh38
rs773328108 Chr.2: 47090346-47090346 on Build GRCh38
rs1038735114 Chr.2: 47090372-47090372 on Build GRCh38
rs954021465 Chr.2: 47090305-47090305 on Build GRCh38
rs765647020 Chr.2: 47090407-47090407 on Build GRCh38
rs376205400 Chr.2: 47090324-47090324 on Build GRCh38
rs765516152 Chr.2: 47090290-47090290 on Build GRCh38
rs563888326 Chr.2: 47090368-47090368 on Build GRCh38
rs745351587 Chr.2: 47090318-47090318 on Build GRCh38
rs961198997 Chr.2: 47090455-47090455 on Build GRCh38
rs541454105 Chr.2: 47090289-47090289 on Build GRCh38
rs760833706 Chr.2: 47090478-47090478 on Build GRCh38
rs772026574 Chr.2: 47090358-47090358 on Build GRCh38
rs774287192 Chr.2: 47090332-47090332 on Build GRCh38
rs759746308 Chr.2: 47090393-47090393 on Build GRCh38
rs192163418 Chr.2: 47090313-47090313 on Build GRCh38
rs1033719751 Chr.2: 47090440-47090440 on Build GRCh38
rs143915542 Chr.2: 47090377-47090377 on Build GRCh38
rs766384005 Chr.2: 47090380-47090380 on Build GRCh38
rs919990451 Chr.2: 47090467-47090467 on Build GRCh38
rs986608570 Chr.2: 47090311-47090313 on Build GRCh38
rs185842543 Chr.2: 47090408-47090408 on Build GRCh38
rs919964999 Chr.2: 47090334-47090334 on Build GRCh38
rs960979716 Chr.2: 47090441-47090441 on Build GRCh38
rs559041331 Chr.2: 47090474-47090474 on Build GRCh38
rs762373142 Chr.2: 47090319-47090319 on Build GRCh38
rs950868468 Chr.2: 47090469-47090469 on Build GRCh38
rs938963875 Chr.2: 47090480-47090480 on Build GRCh38
rs1011337832 Chr.2: 47090398-47090398 on Build GRCh38
rs906088452 Chr.2: 47090345-47090345 on Build GRCh38
rs974883337 Chr.2: 47090490-47090490 on Build GRCh38
rs986703958 Chr.2: 47090447-47090447 on Build GRCh38
rs931292655 Chr.2: 47090335-47090335 on Build GRCh38
rs769460379 Chr.2: 47090437-47090437 on Build GRCh38
rs763409151 Chr.2: 47090420-47090420 on Build GRCh38
rs943031481 Chr.2: 47090362-47090362 on Build GRCh38
rs3739098 Chr.2: 47090492-47090492 on Build GRCh38
rs901496053 Chr.2: 47090374-47090374 on Build GRCh38
rs542301334 Chr.2: 47090322-47090322 on Build GRCh38
rs776840947 Chr.2: 47090385-47090385 on Build GRCh38
rs972805826 Chr.2: 47090320-47090320 on Build GRCh38
rs553087015 Chr.2: 47090306-47090306 on Build GRCh38
rs903519085 Chr.2: 47090431-47090431 on Build GRCh38
rs774671983 Chr.2: 47090398-47090401 on Build GRCh38
rs748892801 Chr.2: 47090323-47090323 on Build GRCh38
rs148206613 Chr.2: 47090468-47090468 on Build GRCh38
rs753210831 Chr.2: 47090299-47090299 on Build GRCh38
rs946178968 Chr.2: 47090493-47090493 on Build GRCh38
rs78092309 Chr.2: 47090450-47090450 on Build GRCh38
rs564835603 Chr.2: 47090421-47090421 on Build GRCh38
rs574392410 Chr.2: 47090488-47090488 on Build GRCh38
rs938865091 Chr.2: 47090373-47090373 on Build GRCh38

View Full Product Details