Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113453129-113453386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADPRHL1
Entrez Gene ID
113622
Gene Name
ADP-ribosylhydrolase like 1
Gene Aliases
ARH2
UniGene
Hs.98669
Gene Chromosome Location
Chr.13: 113421940-113453524 on Build GRCh38
Transcripts
NM_138430.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1365603 c.203C>T p.A68V Chr.13: 113453235-113453235 on Build GRCh38
3399254 c.163G>A p.V55M Chr.13: 113453275-113453275 on Build GRCh38
5473447 c.52G>A p.G18S Chr.13: 113453386-113453386 on Build GRCh38
550787 c.72G>A p.K24K Chr.13: 113453366-113453366 on Build GRCh38
7354620 c.182T>C p.M61T Chr.13: 113453256-113453256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780092180 Chr.13: 113453281-113453281 on Build GRCh38
rs752281449 Chr.13: 113453193-113453193 on Build GRCh38
rs374922964 Chr.13: 113453194-113453194 on Build GRCh38
rs757841091 Chr.13: 113453286-113453286 on Build GRCh38
rs766848804 Chr.13: 113453206-113453206 on Build GRCh38
rs34085933 Chr.13: 113453317-113453317 on Build GRCh38
rs200550427 Chr.13: 113453221-113453221 on Build GRCh38
rs773581591 Chr.13: 113453242-113453242 on Build GRCh38
rs781043297 Chr.13: 113453352-113453352 on Build GRCh38
rs781237593 Chr.13: 113453293-113453293 on Build GRCh38
rs771165404 Chr.13: 113453310-113453310 on Build GRCh38
rs763504678 Chr.13: 113453187-113453187 on Build GRCh38
rs947688222 Chr.13: 113453259-113453259 on Build GRCh38
rs757181816 Chr.13: 113453201-113453201 on Build GRCh38
rs370999543 Chr.13: 113453346-113453346 on Build GRCh38
rs761364646 Chr.13: 113453322-113453322 on Build GRCh38
rs369375515 Chr.13: 113453199-113453199 on Build GRCh38
rs1027077252 Chr.13: 113453156-113453156 on Build GRCh38
rs754251047 Chr.13: 113453182-113453183 on Build GRCh38
rs149499588 Chr.13: 113453275-113453275 on Build GRCh38
rs766088033 Chr.13: 113453347-113453347 on Build GRCh38
rs774697551 Chr.13: 113453253-113453253 on Build GRCh38
rs753510296 Chr.13: 113453200-113453200 on Build GRCh38
rs370894608 Chr.13: 113453191-113453191 on Build GRCh38
rs371511123 Chr.13: 113453179-113453179 on Build GRCh38
rs765006232 Chr.13: 113453323-113453323 on Build GRCh38
rs745589723 Chr.13: 113453205-113453205 on Build GRCh38
rs558010517 Chr.13: 113453294-113453294 on Build GRCh38
rs200853560 Chr.13: 113453280-113453280 on Build GRCh38
rs201411931 Chr.13: 113453207-113453207 on Build GRCh38
rs972963593 Chr.13: 113453157-113453157 on Build GRCh38
rs762019062 Chr.13: 113453239-113453239 on Build GRCh38
rs546248843 Chr.13: 113453292-113453292 on Build GRCh38
rs201345954 Chr.13: 113453240-113453240 on Build GRCh38
rs537275759 Chr.13: 113453318-113453318 on Build GRCh38
rs756572964 Chr.13: 113453175-113453175 on Build GRCh38
rs1053914213 Chr.13: 113453249-113453249 on Build GRCh38
rs747138242 Chr.13: 113453208-113453208 on Build GRCh38
rs753598563 Chr.13: 113453266-113453266 on Build GRCh38
rs754507522 Chr.13: 113453269-113453269 on Build GRCh38
rs750996355 Chr.13: 113453186-113453186 on Build GRCh38
rs141431509 Chr.13: 113453307-113453307 on Build GRCh38
rs928605028 Chr.13: 113453216-113453216 on Build GRCh38
rs138029763 Chr.13: 113453256-113453256 on Build GRCh38
rs756980244 Chr.13: 113453267-113453267 on Build GRCh38
rs35745409 Chr.13: 113453291-113453291 on Build GRCh38
rs776789423 Chr.13: 113453176-113453176 on Build GRCh38
rs778834160 Chr.13: 113453202-113453202 on Build GRCh38
rs776878159 Chr.13: 113453215-113453215 on Build GRCh38
rs377610257 Chr.13: 113453338-113453338 on Build GRCh38
rs567475106 Chr.13: 113453148-113453148 on Build GRCh38
rs770009735 Chr.13: 113453299-113453299 on Build GRCh38
rs746893603 Chr.13: 113453288-113453288 on Build GRCh38
rs777616822 Chr.13: 113453196-113453196 on Build GRCh38
rs765555324 Chr.13: 113453184-113453184 on Build GRCh38
rs935536590 Chr.13: 113453230-113453230 on Build GRCh38
rs994244648 Chr.13: 113453151-113453151 on Build GRCh38
rs766931087 Chr.13: 113453248-113453248 on Build GRCh38
rs750266856 Chr.13: 113453276-113453276 on Build GRCh38
rs952781451 Chr.13: 113453160-113453160 on Build GRCh38
rs140024846 Chr.13: 113453354-113453354 on Build GRCh38
rs748247921 Chr.13: 113453353-113453353 on Build GRCh38
rs148683938 Chr.13: 113453300-113453300 on Build GRCh38
rs375062807 Chr.13: 113453214-113453214 on Build GRCh38
rs774985178 Chr.13: 113453315-113453315 on Build GRCh38
rs369993693 Chr.13: 113453243-113453243 on Build GRCh38

View Full Product Details