Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 138604679-138604904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL22A1
Entrez Gene ID
169044
Gene Name
collagen type XXII alpha 1
Gene Aliases
-
UniGene
Hs.117169
Gene Chromosome Location
Chr.8: 138588235-138914087 on Build GRCh38
Transcripts
NM_152888.2 XM_011516883.2 XM_011516884.2 XM_011516885.2 XM_011516886.2 XM_011516887.2 XM_011516889.2 XM_017013150.1 XM_017013151.1 XM_017013152.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3645616 c.4124G>A p.G1375E Chr.8: 138604750-138604750 on Build GRCh38
4656022 c.4108G>A p.E1370K Chr.8: 138604766-138604766 on Build GRCh38
4546982 c.4123G>A p.G1375R Chr.8: 138604751-138604751 on Build GRCh38
1658899 c.4115G>C p.G1372A Chr.8: 138604759-138604759 on Build GRCh38
4547054 c.4140G>C p.E1380D Chr.8: 138604734-138604734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553570672 Chr.8: 138604699-138604699 on Build GRCh38
rs199727094 Chr.8: 138604719-138604719 on Build GRCh38
rs760633688 Chr.8: 138604797-138604797 on Build GRCh38
rs376144349 Chr.8: 138604795-138604795 on Build GRCh38
rs982113718 Chr.8: 138604833-138604833 on Build GRCh38
rs949079219 Chr.8: 138604741-138604741 on Build GRCh38
rs12546923 Chr.8: 138604803-138604803 on Build GRCh38
rs768832655 Chr.8: 138604820-138604820 on Build GRCh38
rs755682883 Chr.8: 138604871-138604871 on Build GRCh38
rs760045440 Chr.8: 138604848-138604848 on Build GRCh38
rs555863504 Chr.8: 138604789-138604789 on Build GRCh38
rs372510399 Chr.8: 138604732-138604732 on Build GRCh38
rs781348394 Chr.8: 138604706-138604706 on Build GRCh38
rs535442790 Chr.8: 138604841-138604841 on Build GRCh38
rs899219344 Chr.8: 138604879-138604879 on Build GRCh38
rs145150243 Chr.8: 138604863-138604863 on Build GRCh38
rs754606875 Chr.8: 138604711-138604711 on Build GRCh38
rs778717731 Chr.8: 138604716-138604716 on Build GRCh38
rs766262065 Chr.8: 138604800-138604800 on Build GRCh38
rs1037761033 Chr.8: 138604867-138604867 on Build GRCh38
rs371625665 Chr.8: 138604854-138604854 on Build GRCh38
rs527386462 Chr.8: 138604846-138604846 on Build GRCh38
rs117108521 Chr.8: 138604721-138604721 on Build GRCh38
rs745791094 Chr.8: 138604717-138604717 on Build GRCh38
rs753327841 Chr.8: 138604802-138604802 on Build GRCh38
rs377194231 Chr.8: 138604773-138604773 on Build GRCh38
rs541552488 Chr.8: 138604804-138604804 on Build GRCh38
rs773754365 Chr.8: 138604703-138604703 on Build GRCh38
rs138312535 Chr.8: 138604764-138604764 on Build GRCh38
rs776731919 Chr.8: 138604743-138604743 on Build GRCh38
rs765529993 Chr.8: 138604851-138604851 on Build GRCh38
rs769946296 Chr.8: 138604771-138604771 on Build GRCh38
rs763230435 Chr.8: 138604785-138604785 on Build GRCh38
rs1019953446 Chr.8: 138604713-138604713 on Build GRCh38
rs772558806 Chr.8: 138604720-138604720 on Build GRCh38
rs370266282 Chr.8: 138604752-138604752 on Build GRCh38

View Full Product Details