Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33914379-33914652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5O
Entrez Gene ID
539
Gene Name
ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit
Gene Aliases
ATPO, HMC08D05, OSCP
UniGene
Hs.409140
Gene Chromosome Location
Chr.21: 33903453-33915854 on Build GRCh38
Transcripts
NM_001697.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4185669 c.74C>A p.A25D Chr.21: 33914463-33914463 on Build GRCh38
7143161 c.41G>A p.R14Q Chr.21: 33914496-33914496 on Build GRCh38
1728445 c.61G>T p.V21F Chr.21: 33914476-33914476 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs138946175 Chr.21: 33914614-33914614 on Build GRCh38
rs975629649 Chr.21: 33914498-33914498 on Build GRCh38
rs888707454 Chr.21: 33914584-33914584 on Build GRCh38
rs777349814 Chr.21: 33914480-33914480 on Build GRCh38
rs769015276 Chr.21: 33914537-33914537 on Build GRCh38
rs200964053 Chr.21: 33914410-33914410 on Build GRCh38
rs774415235 Chr.21: 33914560-33914560 on Build GRCh38
rs773624767 Chr.21: 33914427-33914427 on Build GRCh38
rs12626438 Chr.21: 33914436-33914436 on Build GRCh38
rs748506916 Chr.21: 33914546-33914546 on Build GRCh38
rs143025513 Chr.21: 33914565-33914565 on Build GRCh38
rs1038109817 Chr.21: 33914585-33914585 on Build GRCh38
rs943481003 Chr.21: 33914481-33914481 on Build GRCh38
rs533283808 Chr.21: 33914572-33914573 on Build GRCh38
rs755961843 Chr.21: 33914467-33914467 on Build GRCh38
rs752455954 Chr.21: 33914461-33914461 on Build GRCh38
rs765548655 Chr.21: 33914421-33914421 on Build GRCh38
rs533468498 Chr.21: 33914592-33914592 on Build GRCh38
rs761317773 Chr.21: 33914415-33914415 on Build GRCh38
rs763064697 Chr.21: 33914430-33914430 on Build GRCh38
rs777347186 Chr.21: 33914544-33914544 on Build GRCh38
rs762311058 Chr.21: 33914413-33914413 on Build GRCh38
rs531517223 Chr.21: 33914567-33914567 on Build GRCh38
rs374587852 Chr.21: 33914453-33914453 on Build GRCh38
rs776839804 Chr.21: 33914412-33914412 on Build GRCh38
rs745437519 Chr.21: 33914522-33914522 on Build GRCh38
rs144166454 Chr.21: 33914478-33914478 on Build GRCh38
rs542905799 Chr.21: 33914499-33914499 on Build GRCh38
rs764361907 Chr.21: 33914531-33914532 on Build GRCh38
rs190343471 Chr.21: 33914497-33914497 on Build GRCh38
rs193208543 Chr.21: 33914579-33914579 on Build GRCh38
rs751473882 Chr.21: 33914445-33914445 on Build GRCh38
rs770443406 Chr.21: 33914409-33914409 on Build GRCh38
rs201086460 Chr.21: 33914405-33914405 on Build GRCh38
rs7281423 Chr.21: 33914590-33914590 on Build GRCh38
rs767538170 Chr.21: 33914459-33914459 on Build GRCh38
rs1023521763 Chr.21: 33914607-33914607 on Build GRCh38
rs748943333 Chr.21: 33914493-33914493 on Build GRCh38

View Full Product Details