Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93271087-93271348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS50
Entrez Gene ID
55610
Gene Name
VPS50, EARP/GARPII complex subunit
Gene Aliases
CCDC132, VPS54L
UniGene
Hs.202424
Gene Chromosome Location
Chr.7: 93232340-93361123 on Build GRCh38
Transcripts
NM_001257998.1 NM_017667.3 NM_024553.2 XM_011516395.2 XM_011516396.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2864701 c.660-5A>G p.? Chr.7: 93271215-93271215 on Build GRCh38
2864702 c.660-5A>G p.? Chr.7: 93271215-93271215 on Build GRCh38
1635490 c.693A>G p.E231E Chr.7: 93271253-93271253 on Build GRCh38
2864703 c.700G>C p.E234Q Chr.7: 93271260-93271260 on Build GRCh38
2864704 c.700G>C p.E234Q Chr.7: 93271260-93271260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561311041 Chr.7: 93271164-93271164 on Build GRCh38
rs759563759 Chr.7: 93271201-93271201 on Build GRCh38
rs750985350 Chr.7: 93271201-93271201 on Build GRCh38
rs375400119 Chr.7: 93271244-93271244 on Build GRCh38
rs756963351 Chr.7: 93271201-93271202 on Build GRCh38
rs767795416 Chr.7: 93271203-93271203 on Build GRCh38
rs749911412 Chr.7: 93271265-93271265 on Build GRCh38
rs758204321 Chr.7: 93271201-93271204 on Build GRCh38
rs1033691452 Chr.7: 93271118-93271118 on Build GRCh38
rs1022991353 Chr.7: 93271250-93271250 on Build GRCh38
rs995235919 Chr.7: 93271321-93271321 on Build GRCh38
rs769728856 Chr.7: 93271309-93271309 on Build GRCh38
rs530819638 Chr.7: 93271200-93271201 on Build GRCh38
rs993176284 Chr.7: 93271162-93271162 on Build GRCh38
rs377632505 Chr.7: 93271200-93271200 on Build GRCh38
rs765219386 Chr.7: 93271199-93271200 on Build GRCh38
rs781087648 Chr.7: 93271293-93271293 on Build GRCh38
rs754754612 Chr.7: 93271276-93271276 on Build GRCh38
rs934243214 Chr.7: 93271272-93271272 on Build GRCh38
rs776454586 Chr.7: 93271190-93271191 on Build GRCh38
rs62466775 Chr.7: 93271216-93271216 on Build GRCh38
rs764623180 Chr.7: 93271235-93271235 on Build GRCh38
rs201670312 Chr.7: 93271266-93271266 on Build GRCh38
rs527426642 Chr.7: 93271173-93271173 on Build GRCh38
rs746729626 Chr.7: 93271271-93271271 on Build GRCh38
rs746280864 Chr.7: 93271203-93271204 on Build GRCh38
rs908504264 Chr.7: 93271140-93271140 on Build GRCh38
rs12112676 Chr.7: 93271204-93271204 on Build GRCh38
rs932410135 Chr.7: 93271199-93271199 on Build GRCh38
rs767597992 Chr.7: 93271190-93271190 on Build GRCh38
rs187415569 Chr.7: 93271124-93271124 on Build GRCh38
rs1014486612 Chr.7: 93271249-93271249 on Build GRCh38
rs59890424 Chr.7: 93271214-93271214 on Build GRCh38
rs533562042 Chr.7: 93271215-93271215 on Build GRCh38
rs765810206 Chr.7: 93271268-93271268 on Build GRCh38
rs547444423 Chr.7: 93271200-93271200 on Build GRCh38
rs757835008 Chr.7: 93271226-93271226 on Build GRCh38
rs761057931 Chr.7: 93271205-93271205 on Build GRCh38
rs759306553 Chr.7: 93271191-93271191 on Build GRCh38
rs976393853 Chr.7: 93271169-93271172 on Build GRCh38
rs917088696 Chr.7: 93271198-93271198 on Build GRCh38
rs1053240086 Chr.7: 93271211-93271211 on Build GRCh38
rs754289370 Chr.7: 93271217-93271217 on Build GRCh38
rs76003785 Chr.7: 93271214-93271214 on Build GRCh38
rs748003334 Chr.7: 93271304-93271304 on Build GRCh38
rs958633650 Chr.7: 93271119-93271119 on Build GRCh38
rs759241361 Chr.7: 93271134-93271134 on Build GRCh38
rs756532248 Chr.7: 93271271-93271273 on Build GRCh38

View Full Product Details