Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 76355267-76355492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP4
Entrez Gene ID
85445
Gene Name
contactin associated protein like 4
Gene Aliases
CASPR4
UniGene
Hs.461389
Gene Chromosome Location
Chr.16: 76277217-76563610 on Build GRCh38
Transcripts
NM_001322178.1 NM_001322179.1 NM_001322180.1 NM_001322181.1 NM_001322187.1 NM_001322188.1 NM_001322189.1 NM_001322190.1 NM_001322191.1 NM_033401.4 NM_138994.4 NR_136210.1 NR_136213.1 NR_136216.1 XM_011523403.2 XM_017023805.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3512325 c.200C>T p.T67I Chr.16: 76355396-76355396 on Build GRCh38
382683 c.283A>G p.T95A Chr.16: 76355407-76355407 on Build GRCh38
5920965 c.122G>A p.G41E Chr.16: 76355318-76355318 on Build GRCh38
6247631 c.168G>T p.L56F Chr.16: 76355364-76355364 on Build GRCh38
7361858 c.277T>A p.W93R Chr.16: 76355473-76355473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772544918 Chr.16: 76355391-76355391 on Build GRCh38
rs775287770 Chr.16: 76355427-76355427 on Build GRCh38
rs781538603 Chr.16: 76355406-76355406 on Build GRCh38
rs1029547896 Chr.16: 76355469-76355469 on Build GRCh38
rs150850793 Chr.16: 76355382-76355382 on Build GRCh38
rs143795961 Chr.16: 76355443-76355443 on Build GRCh38
rs753898393 Chr.16: 76355449-76355449 on Build GRCh38
rs780177745 Chr.16: 76355361-76355361 on Build GRCh38
rs763916503 Chr.16: 76355438-76355438 on Build GRCh38
rs748480259 Chr.16: 76355387-76355387 on Build GRCh38
rs773373872 Chr.16: 76355442-76355442 on Build GRCh38
rs1026484587 Chr.16: 76355368-76355368 on Build GRCh38
rs572403674 Chr.16: 76355306-76355306 on Build GRCh38
rs760154699 Chr.16: 76355301-76355301 on Build GRCh38
rs138220882 Chr.16: 76355397-76355397 on Build GRCh38
rs777332448 Chr.16: 76355396-76355396 on Build GRCh38
rs762784908 Chr.16: 76355432-76355432 on Build GRCh38
rs796632029 Chr.16: 76355341-76355341 on Build GRCh38
rs964702992 Chr.16: 76355454-76355454 on Build GRCh38
rs374909736 Chr.16: 76355398-76355398 on Build GRCh38
rs202210201 Chr.16: 76355459-76355459 on Build GRCh38
rs147250119 Chr.16: 76355451-76355451 on Build GRCh38
rs772806876 Chr.16: 76355299-76355299 on Build GRCh38
rs371784566 Chr.16: 76355337-76355337 on Build GRCh38
rs141544092 Chr.16: 76355323-76355323 on Build GRCh38
rs371066872 Chr.16: 76355333-76355333 on Build GRCh38
rs756188053 Chr.16: 76355352-76355352 on Build GRCh38
rs146643417 Chr.16: 76355302-76355302 on Build GRCh38
rs532410036 Chr.16: 76355384-76355384 on Build GRCh38
rs750634133 Chr.16: 76355349-76355349 on Build GRCh38
rs769681418 Chr.16: 76355417-76355417 on Build GRCh38
rs191607702 Chr.16: 76355313-76355313 on Build GRCh38
rs754356758 Chr.16: 76355378-76355378 on Build GRCh38
rs971627059 Chr.16: 76355312-76355312 on Build GRCh38
rs767676346 Chr.16: 76355340-76355340 on Build GRCh38
rs759474348 Chr.16: 76355404-76355404 on Build GRCh38
rs759460015 Chr.16: 76355309-76355309 on Build GRCh38

View Full Product Details