Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23467509-23467782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGA2
Entrez Gene ID
23062
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 2
Gene Aliases
VEAR
UniGene
Hs.460336
Gene Chromosome Location
Chr.16: 23463542-23510494 on Build GRCh38
Transcripts
NM_015044.4 XM_017023074.1 XM_017023075.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6839011 c.1756C>A p.L586M Chr.16: 23467676-23467676 on Build GRCh38
293742 c.1796_1798delGAG p.G599delG Chr.16: 23467634-23467636 on Build GRCh38
5850493 c.1781C>T p.P594L Chr.16: 23467651-23467651 on Build GRCh38
3817563 c.1773T>G p.G591G Chr.16: 23467659-23467659 on Build GRCh38
5772678 c.1807G>C p.D603H Chr.16: 23467625-23467625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779834299 Chr.16: 23467643-23467643 on Build GRCh38
rs746184190 Chr.16: 23467677-23467677 on Build GRCh38
rs373329210 Chr.16: 23467728-23467728 on Build GRCh38
rs761387519 Chr.16: 23467620-23467620 on Build GRCh38
rs749739332 Chr.16: 23467588-23467588 on Build GRCh38
rs934499580 Chr.16: 23467727-23467727 on Build GRCh38
rs926385320 Chr.16: 23467754-23467754 on Build GRCh38
rs1052857845 Chr.16: 23467549-23467549 on Build GRCh38
rs148356247 Chr.16: 23467657-23467657 on Build GRCh38
rs201587696 Chr.16: 23467609-23467609 on Build GRCh38
rs199705736 Chr.16: 23467604-23467604 on Build GRCh38
rs200722860 Chr.16: 23467715-23467715 on Build GRCh38
rs185467966 Chr.16: 23467690-23467690 on Build GRCh38
rs764765850 Chr.16: 23467704-23467704 on Build GRCh38
rs1011671025 Chr.16: 23467574-23467574 on Build GRCh38
rs768157448 Chr.16: 23467688-23467688 on Build GRCh38
rs777854676 Chr.16: 23467559-23467559 on Build GRCh38
rs550362454 Chr.16: 23467599-23467599 on Build GRCh38
rs370266306 Chr.16: 23467627-23467627 on Build GRCh38
rs778130677 Chr.16: 23467575-23467575 on Build GRCh38
rs779086522 Chr.16: 23467703-23467703 on Build GRCh38
rs968304516 Chr.16: 23467589-23467589 on Build GRCh38
rs372657289 Chr.16: 23467552-23467552 on Build GRCh38
rs753924908 Chr.16: 23467752-23467752 on Build GRCh38
rs1007847798 Chr.16: 23467603-23467603 on Build GRCh38
rs781364790 Chr.16: 23467548-23467548 on Build GRCh38
rs980535056 Chr.16: 23467753-23467753 on Build GRCh38
rs763793027 Chr.16: 23467611-23467611 on Build GRCh38
rs760106544 Chr.16: 23467600-23467600 on Build GRCh38
rs761476497 Chr.16: 23467694-23467694 on Build GRCh38
rs764295239 Chr.16: 23467744-23467744 on Build GRCh38
rs754908936 Chr.16: 23467738-23467738 on Build GRCh38
rs750145020 Chr.16: 23467634-23467634 on Build GRCh38
rs142107518 Chr.16: 23467668-23467668 on Build GRCh38
rs948723046 Chr.16: 23467691-23467691 on Build GRCh38
rs758401729 Chr.16: 23467639-23467639 on Build GRCh38
rs770264422 Chr.16: 23467541-23467541 on Build GRCh38
rs756260632 Chr.16: 23467648-23467648 on Build GRCh38
rs750283251 Chr.16: 23467714-23467714 on Build GRCh38
rs752882262 Chr.16: 23467644-23467644 on Build GRCh38
rs758210704 Chr.16: 23467546-23467546 on Build GRCh38
rs746297665 Chr.16: 23467717-23467719 on Build GRCh38
rs749721440 Chr.16: 23467634-23467636 on Build GRCh38
rs1052912412 Chr.16: 23467756-23467756 on Build GRCh38
rs779362062 Chr.16: 23467671-23467671 on Build GRCh38
rs568191925 Chr.16: 23467707-23467707 on Build GRCh38
rs909361176 Chr.16: 23467695-23467695 on Build GRCh38
rs757783939 Chr.16: 23467681-23467682 on Build GRCh38
rs772681875 Chr.16: 23467684-23467684 on Build GRCh38
rs775844878 Chr.16: 23467685-23467685 on Build GRCh38
rs985265053 Chr.16: 23467689-23467689 on Build GRCh38
rs777935824 Chr.16: 23467656-23467656 on Build GRCh38
rs748146470 Chr.16: 23467550-23467550 on Build GRCh38
rs1028017846 Chr.16: 23467582-23467582 on Build GRCh38
rs753484122 Chr.16: 23467619-23467619 on Build GRCh38
rs771430125 Chr.16: 23467598-23467598 on Build GRCh38
rs1017669430 Chr.16: 23467616-23467616 on Build GRCh38

View Full Product Details