Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111499874-111500133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADORA3
Entrez Gene ID
140
Gene Name
adenosine A3 receptor
Gene Aliases
A3AR
UniGene
Hs.281342
Gene Chromosome Location
Chr.1: 111499429-111504121 on Build GRCh38
Transcripts
NM_000677.3 NM_001302678.1 NM_001302679.1 NM_001081976.2 NM_001302680.1 NM_020683.6

Gene Symbol
TMIGD3
Entrez Gene ID
57413
Gene Name
transmembrane and immunoglobulin domain containing 3
Gene Aliases
AD026
UniGene
Hs.281342
Gene Chromosome Location
Chr.1: 111483348-111564003 on Build GRCh38
Transcripts
NM_000677.3 NM_001302678.1 NM_001302679.1 NM_001081976.2 NM_001302680.1 NM_020683.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4393205 c.895G>T p.A299S Chr.1: 111500012-111500012 on Build GRCh38
5629280 c.836C>G p.P279R Chr.1: 111500071-111500071 on Build GRCh38
3360220 c.936C>T p.S312S Chr.1: 111499971-111499971 on Build GRCh38
1244523 c.808C>A p.L270M Chr.1: 111500099-111500099 on Build GRCh38
5653206 c.945G>T p.K315N Chr.1: 111499962-111499962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139193735 Chr.1: 111500066-111500066 on Build GRCh38
rs897809219 Chr.1: 111499918-111499918 on Build GRCh38
rs367610688 Chr.1: 111499934-111499934 on Build GRCh38
rs772854601 Chr.1: 111499966-111499966 on Build GRCh38
rs561203407 Chr.1: 111499988-111499988 on Build GRCh38
rs770697740 Chr.1: 111500031-111500031 on Build GRCh38
rs754410734 Chr.1: 111500089-111500089 on Build GRCh38
rs755109360 Chr.1: 111500013-111500013 on Build GRCh38
rs147238368 Chr.1: 111500073-111500073 on Build GRCh38
rs776719047 Chr.1: 111500040-111500040 on Build GRCh38
rs745783278 Chr.1: 111500042-111500042 on Build GRCh38
rs781394667 Chr.1: 111499922-111499922 on Build GRCh38
rs2229155 Chr.1: 111500010-111500010 on Build GRCh38
rs750730029 Chr.1: 111500008-111500008 on Build GRCh38
rs778961251 Chr.1: 111500020-111500020 on Build GRCh38
rs775106794 Chr.1: 111500062-111500062 on Build GRCh38
rs760952472 Chr.1: 111500067-111500067 on Build GRCh38
rs757705456 Chr.1: 111499919-111499919 on Build GRCh38
rs983223023 Chr.1: 111499930-111499930 on Build GRCh38
rs758461041 Chr.1: 111500112-111500112 on Build GRCh38
rs765352303 Chr.1: 111500107-111500107 on Build GRCh38
rs759126309 Chr.1: 111499981-111499981 on Build GRCh38
rs770799551 Chr.1: 111499975-111499975 on Build GRCh38
rs746814002 Chr.1: 111500030-111500030 on Build GRCh38
rs775118942 Chr.1: 111500088-111500088 on Build GRCh38
rs763675616 Chr.1: 111500065-111500065 on Build GRCh38
rs371299718 Chr.1: 111499939-111499939 on Build GRCh38
rs544398617 Chr.1: 111499968-111499968 on Build GRCh38
rs752452241 Chr.1: 111499989-111499989 on Build GRCh38
rs2800889 Chr.1: 111500110-111500110 on Build GRCh38
rs745764662 Chr.1: 111499961-111499961 on Build GRCh38
rs201053017 Chr.1: 111499952-111499952 on Build GRCh38
rs748411636 Chr.1: 111500021-111500021 on Build GRCh38
rs755448584 Chr.1: 111500105-111500105 on Build GRCh38
rs752327361 Chr.1: 111499905-111499905 on Build GRCh38
rs765016750 Chr.1: 111499904-111499904 on Build GRCh38
rs751003117 Chr.1: 111499924-111499924 on Build GRCh38
rs768682866 Chr.1: 111499945-111499945 on Build GRCh38
rs529928933 Chr.1: 111499993-111499993 on Build GRCh38
rs778005823 Chr.1: 111500029-111500029 on Build GRCh38
rs1040252885 Chr.1: 111500102-111500102 on Build GRCh38
rs771971217 Chr.1: 111499962-111499962 on Build GRCh38
rs756545764 Chr.1: 111499925-111499925 on Build GRCh38
rs767963822 Chr.1: 111499999-111499999 on Build GRCh38
rs17852706 Chr.1: 111500025-111500025 on Build GRCh38
rs769492097 Chr.1: 111500052-111500052 on Build GRCh38
rs762713981 Chr.1: 111500064-111500064 on Build GRCh38
rs528585080 Chr.1: 111500080-111500080 on Build GRCh38
rs368754980 Chr.1: 111499976-111499976 on Build GRCh38
rs770181778 Chr.1: 111499982-111499982 on Build GRCh38
rs766825659 Chr.1: 111500009-111500009 on Build GRCh38
rs758529872 Chr.1: 111500027-111500027 on Build GRCh38

View Full Product Details