Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201145345-201145600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFLAR
Entrez Gene ID
8837
Gene Name
CASP8 and FADD like apoptosis regulator
Gene Aliases
CASH, CASP8AP1, CLARP, Casper, FLAME, FLAME-1, FLAME1, FLIP, I-FLICE, MRIT, c-FLIP, c-FLIPL, c-FLIPR, c-FLIPS
UniGene
Hs.390736
Gene Chromosome Location
Chr.2: 201116015-201172688 on Build GRCh38
Transcripts
NM_001127183.2 NM_001202515.1 NM_001202516.1 NM_001202517.1 NM_001202518.1 NM_001202519.1 NM_001308042.1 NM_003879.5 XM_017005189.1 XM_017005190.1 XM_017005191.1 XM_017005192.1 XM_017005193.1 XM_017005194.1 XM_017005195.1 XM_017005196.1 NR_040030.1

Gene Symbol
CFLAR-AS1
Entrez Gene ID
65072
Gene Name
CFLAR antisense RNA 1
Gene Aliases
ALS2CR10
UniGene
Hs.664613
Gene Chromosome Location
Chr.2: 201140289-201157792 on Build GRCh38
Transcripts
NM_001127183.2 NM_001202515.1 NM_001202516.1 NM_001202517.1 NM_001202518.1 NM_001202519.1 NM_001308042.1 NM_003879.5 XM_017005189.1 XM_017005190.1 XM_017005191.1 XM_017005192.1 XM_017005193.1 XM_017005194.1 XM_017005195.1 XM_017005196.1 NR_040030.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5973534 c.640A>G p.K214E Chr.2: 201145411-201145411 on Build GRCh38
168709 c.653G>A p.G218D Chr.2: 201145424-201145424 on Build GRCh38
6366538 c.633A>T p.Q211H Chr.2: 201145404-201145404 on Build GRCh38
6627571 c.661+1G>A p.? Chr.2: 201145433-201145433 on Build GRCh38
6323764 c.656C>T p.A219V Chr.2: 201145427-201145427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769477054 Chr.2: 201145425-201145425 on Build GRCh38
rs575079661 Chr.2: 201145503-201145503 on Build GRCh38
rs764442521 Chr.2: 201145458-201145458 on Build GRCh38
rs777136234 Chr.2: 201145399-201145399 on Build GRCh38
rs558234044 Chr.2: 201145450-201145450 on Build GRCh38
rs746592349 Chr.2: 201145404-201145404 on Build GRCh38
rs552264985 Chr.2: 201145430-201145430 on Build GRCh38
rs570493276 Chr.2: 201145482-201145482 on Build GRCh38
rs906478759 Chr.2: 201145403-201145403 on Build GRCh38
rs952078880 Chr.2: 201145572-201145572 on Build GRCh38
rs972184858 Chr.2: 201145565-201145565 on Build GRCh38
rs556697165 Chr.2: 201145496-201145496 on Build GRCh38
rs984856224 Chr.2: 201145573-201145573 on Build GRCh38
rs369605425 Chr.2: 201145483-201145483 on Build GRCh38
rs1019184783 Chr.2: 201145529-201145529 on Build GRCh38
rs774263004 Chr.2: 201145461-201145461 on Build GRCh38
rs966289181 Chr.2: 201145554-201145554 on Build GRCh38
rs771553744 Chr.2: 201145528-201145528 on Build GRCh38
rs1044988877 Chr.2: 201145384-201145384 on Build GRCh38
rs771556544 Chr.2: 201145396-201145396 on Build GRCh38
rs986239508 Chr.2: 201145517-201145517 on Build GRCh38
rs1030795286 Chr.2: 201145422-201145422 on Build GRCh38
rs774043719 Chr.2: 201145443-201145444 on Build GRCh38
rs768088440 Chr.2: 201145462-201145462 on Build GRCh38
rs3769829 Chr.2: 201145410-201145410 on Build GRCh38
rs745543569 Chr.2: 201145466-201145466 on Build GRCh38
rs776568972 Chr.2: 201145415-201145415 on Build GRCh38
rs3769828 Chr.2: 201145567-201145567 on Build GRCh38
rs376941002 Chr.2: 201145438-201145438 on Build GRCh38
rs538090713 Chr.2: 201145494-201145494 on Build GRCh38
rs770435962 Chr.2: 201145438-201145439 on Build GRCh38
rs533504297 Chr.2: 201145440-201145440 on Build GRCh38
rs566120564 Chr.2: 201145420-201145420 on Build GRCh38
rs747763801 Chr.2: 201145386-201145386 on Build GRCh38
rs988075161 Chr.2: 201145535-201145535 on Build GRCh38
rs935438381 Chr.2: 201145532-201145532 on Build GRCh38
rs759180946 Chr.2: 201145454-201145457 on Build GRCh38
rs13424615 Chr.2: 201145378-201145378 on Build GRCh38

View Full Product Details