Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151797290-151797515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G3BP1
Entrez Gene ID
10146
Gene Name
G3BP stress granule assembly factor 1
Gene Aliases
G3BP, HDH-VIII
UniGene
Hs.3353
Gene Chromosome Location
Chr.5: 151771915-151811352 on Build GRCh38
Transcripts
NM_005754.2 NM_198395.1 XM_006714749.3 XM_006714750.3 XM_017008922.1 XM_017008923.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1064649 c.652G>A p.V218I Chr.5: 151797339-151797339 on Build GRCh38
6410265 c.673G>A p.E225K Chr.5: 151797360-151797360 on Build GRCh38
109611 c.670C>T p.P224S Chr.5: 151797357-151797357 on Build GRCh38
4602804 c.634G>T p.E212* Chr.5: 151797321-151797321 on Build GRCh38
3721871 c.631C>T p.Q211* Chr.5: 151797318-151797318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902774439 Chr.5: 151797333-151797333 on Build GRCh38
rs148167548 Chr.5: 151797317-151797317 on Build GRCh38
rs765658988 Chr.5: 151797337-151797337 on Build GRCh38
rs548320196 Chr.5: 151797403-151797403 on Build GRCh38
rs754302303 Chr.5: 151797478-151797483 on Build GRCh38
rs760344048 Chr.5: 151797446-151797446 on Build GRCh38
rs759116158 Chr.5: 151797325-151797325 on Build GRCh38
rs768409286 Chr.5: 151797454-151797454 on Build GRCh38
rs558552988 Chr.5: 151797464-151797464 on Build GRCh38
rs781195002 Chr.5: 151797399-151797399 on Build GRCh38
rs201027869 Chr.5: 151797327-151797327 on Build GRCh38
rs147296160 Chr.5: 151797338-151797338 on Build GRCh38
rs370777826 Chr.5: 151797438-151797438 on Build GRCh38
rs1000158539 Chr.5: 151797334-151797334 on Build GRCh38
rs755058108 Chr.5: 151797364-151797364 on Build GRCh38
rs370787939 Chr.5: 151797467-151797467 on Build GRCh38
rs537765817 Chr.5: 151797474-151797474 on Build GRCh38
rs1023381854 Chr.5: 151797355-151797355 on Build GRCh38
rs748217398 Chr.5: 151797400-151797400 on Build GRCh38
rs181562129 Chr.5: 151797475-151797475 on Build GRCh38
rs761376769 Chr.5: 151797459-151797459 on Build GRCh38
rs770937197 Chr.5: 151797402-151797402 on Build GRCh38
rs780347568 Chr.5: 151797351-151797351 on Build GRCh38
rs974498723 Chr.5: 151797341-151797341 on Build GRCh38
rs775211854 Chr.5: 151797445-151797445 on Build GRCh38
rs763408429 Chr.5: 151797468-151797468 on Build GRCh38
rs143045415 Chr.5: 151797357-151797357 on Build GRCh38
rs780577312 Chr.5: 151797467-151797468 on Build GRCh38
rs1051898034 Chr.5: 151797485-151797485 on Build GRCh38
rs1046466289 Chr.5: 151797484-151797484 on Build GRCh38
rs752819456 Chr.5: 151797480-151797480 on Build GRCh38
rs970588999 Chr.5: 151797369-151797369 on Build GRCh38
rs142032366 Chr.5: 151797336-151797336 on Build GRCh38
rs756479611 Chr.5: 151797377-151797379 on Build GRCh38
rs376309064 Chr.5: 151797322-151797322 on Build GRCh38
rs776345653 Chr.5: 151797457-151797457 on Build GRCh38
rs112027688 Chr.5: 151797410-151797410 on Build GRCh38
rs764902207 Chr.5: 151797437-151797437 on Build GRCh38
rs78729597 Chr.5: 151797476-151797476 on Build GRCh38
rs757895524 Chr.5: 151797335-151797335 on Build GRCh38
rs1012028655 Chr.5: 151797339-151797339 on Build GRCh38
rs760291090 Chr.5: 151797321-151797321 on Build GRCh38
rs933375033 Chr.5: 151797477-151797477 on Build GRCh38
rs754410160 Chr.5: 151797331-151797331 on Build GRCh38
rs751743912 Chr.5: 151797358-151797358 on Build GRCh38

View Full Product Details