Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 32456170-32456417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC7
Entrez Gene ID
79741
Gene Name
coiled-coil domain containing 7
Gene Aliases
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
UniGene
Hs.585464
Gene Chromosome Location
Chr.10: 32446082-32882864 on Build GRCh38
Transcripts
NM_001026383.2 NM_001321115.1 NM_145023.5 NR_109826.1 NR_109827.1 XM_006717505.1 XM_006717506.1 XM_011519663.1 XM_011519664.1 XM_011519666.1 XM_011519667.1 XM_011519668.1 XM_011519669.1 XM_011519670.1 XM_011519671.1 XM_011519672.1 XM_011519673.1 XM_011519674.1 XM_011519675.1 XM_011519676.1 XM_011519677.1 XM_011519678.1 XM_011519679.1 XM_011519684.2 XM_011519686.2 XM_011519688.2 XM_017016638.1 XM_017016639.1 XM_017016640.1 XM_017016643.1 XM_017016644.1 XM_017016645.1 XM_017016647.1 XM_017016650.1 XM_017016651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3675712 c.456T>A p.N152K Chr.10: 32456334-32456334 on Build GRCh38
3807079 c.424C>T p.Q142* Chr.10: 32456302-32456302 on Build GRCh38
3751739 c.456T>C p.N152N Chr.10: 32456334-32456334 on Build GRCh38
3751738 c.442A>C p.K148Q Chr.10: 32456320-32456320 on Build GRCh38
6622385 c.410C>T p.S137L Chr.10: 32456288-32456288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749846682 Chr.10: 32456203-32456203 on Build GRCh38
rs772802509 Chr.10: 32456337-32456337 on Build GRCh38
rs749124265 Chr.10: 32456338-32456338 on Build GRCh38
rs376373220 Chr.10: 32456264-32456264 on Build GRCh38
rs777946513 Chr.10: 32456309-32456309 on Build GRCh38
rs776052180 Chr.10: 32456227-32456227 on Build GRCh38
rs146377189 Chr.10: 32456312-32456312 on Build GRCh38
rs1030835450 Chr.10: 32456238-32456238 on Build GRCh38
rs372040541 Chr.10: 32456370-32456370 on Build GRCh38
rs141076922 Chr.10: 32456346-32456346 on Build GRCh38
rs371686762 Chr.10: 32456320-32456322 on Build GRCh38
rs764027763 Chr.10: 32456383-32456383 on Build GRCh38
rs779088429 Chr.10: 32456322-32456322 on Build GRCh38
rs989727242 Chr.10: 32456296-32456296 on Build GRCh38
rs376733998 Chr.10: 32456275-32456275 on Build GRCh38
rs761305863 Chr.10: 32456236-32456236 on Build GRCh38
rs766040932 Chr.10: 32456279-32456279 on Build GRCh38
rs747840685 Chr.10: 32456333-32456333 on Build GRCh38
rs56391924 Chr.10: 32456320-32456320 on Build GRCh38
rs756237974 Chr.10: 32456307-32456307 on Build GRCh38
rs1048703228 Chr.10: 32456349-32456349 on Build GRCh38
rs746612707 Chr.10: 32456217-32456217 on Build GRCh38
rs544334165 Chr.10: 32456369-32456369 on Build GRCh38
rs768310384 Chr.10: 32456220-32456220 on Build GRCh38
rs370748954 Chr.10: 32456282-32456282 on Build GRCh38
rs7897978 Chr.10: 32456334-32456334 on Build GRCh38
rs772993519 Chr.10: 32456254-32456254 on Build GRCh38
rs779362002 Chr.10: 32456212-32456212 on Build GRCh38
rs139970259 Chr.10: 32456387-32456387 on Build GRCh38
rs769396847 Chr.10: 32456240-32456240 on Build GRCh38
rs914104143 Chr.10: 32456306-32456306 on Build GRCh38
rs142093528 Chr.10: 32456289-32456289 on Build GRCh38
rs369861851 Chr.10: 32456205-32456205 on Build GRCh38
rs113888697 Chr.10: 32456288-32456288 on Build GRCh38
rs370129971 Chr.10: 32456328-32456328 on Build GRCh38
rs184849393 Chr.10: 32456367-32456367 on Build GRCh38
rs759770084 Chr.10: 32456284-32456284 on Build GRCh38
rs746235004 Chr.10: 32456325-32456325 on Build GRCh38
rs907398294 Chr.10: 32456207-32456207 on Build GRCh38
rs1002955904 Chr.10: 32456210-32456210 on Build GRCh38
rs760587057 Chr.10: 32456393-32456394 on Build GRCh38
rs759394979 Chr.10: 32456350-32456350 on Build GRCh38
rs967980208 Chr.10: 32456301-32456301 on Build GRCh38
rs757638109 Chr.10: 32456394-32456394 on Build GRCh38
rs753961326 Chr.10: 32456386-32456386 on Build GRCh38
rs373674730 Chr.10: 32456330-32456330 on Build GRCh38

View Full Product Details