Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97124531-97124782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36
Entrez Gene ID
375248
Gene Name
ankyrin repeat domain 36
Gene Aliases
UNQ2430
UniGene
Hs.541894
Gene Chromosome Location
Chr.2: 97113186-97264521 on Build GRCh38
Transcripts
NM_001164315.1 XM_006712514.3 XM_011511130.1 XM_011511131.1 XM_017004009.1 XM_017004010.1 XM_017004011.1 XM_017004012.1 XM_017004013.1 XM_017004014.1 XM_017004015.1 XM_017004016.1 XM_017004017.1 XM_017004018.1 XM_017004019.1 XM_017004020.1 XM_017004021.1 XM_017004022.1 XM_017004023.1 XM_017004024.1 XM_017004025.1 XM_017004026.1 XM_017004027.1 XM_017004028.1 XM_017004029.1 XM_017004030.1 XM_017004031.1 XM_017004032.1 XM_017004033.1 XM_017004034.1 XM_017004035.1 XM_017004036.1 XM_017004037.1 XM_017004038.1 XM_017004039.1 XM_017004040.1 XM_017004041.1 XM_017004042.1 XM_017004043.1 XM_017004044.1 XM_017004045.1 XM_017004046.1 XM_017004047.1 XM_017004048.1 XM_017004049.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5004071 c.680A>T p.D227V Chr.2: 97124546-97124546 on Build GRCh38
6419979 c.698C>T p.A233V Chr.2: 97124564-97124564 on Build GRCh38
5004072 c.680A>T p.D227V Chr.2: 97124546-97124546 on Build GRCh38
6419980 c.698C>T p.A233V Chr.2: 97124564-97124564 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781284967 Chr.2: 97124562-97124562 on Build GRCh38
rs1017687421 Chr.2: 97124655-97124655 on Build GRCh38
rs770933893 Chr.2: 97124555-97124555 on Build GRCh38
rs4491760 Chr.2: 97124693-97124693 on Build GRCh38
rs774097401 Chr.2: 97124621-97124621 on Build GRCh38
rs1007689268 Chr.2: 97124649-97124649 on Build GRCh38
rs567976661 Chr.2: 97124685-97124685 on Build GRCh38
rs534114407 Chr.2: 97124742-97124742 on Build GRCh38
rs763159129 Chr.2: 97124554-97124554 on Build GRCh38
rs113015045 Chr.2: 97124661-97124661 on Build GRCh38
rs893360545 Chr.2: 97124611-97124611 on Build GRCh38
rs548013553 Chr.2: 97124591-97124591 on Build GRCh38
rs775234250 Chr.2: 97124587-97124587 on Build GRCh38
rs770051110 Chr.2: 97124584-97124584 on Build GRCh38
rs768626361 Chr.2: 97124609-97124609 on Build GRCh38
rs746044379 Chr.2: 97124575-97124575 on Build GRCh38
rs4502463 Chr.2: 97124589-97124589 on Build GRCh38
rs1026384177 Chr.2: 97124753-97124753 on Build GRCh38

View Full Product Details