Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2120264-2120489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1C
Entrez Gene ID
775
Gene Name
calcium voltage-gated channel subunit alpha1 C
Gene Aliases
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS
UniGene
Hs.118262
Gene Chromosome Location
Chr.12: 1969677-2697949 on Build GRCh38
Transcripts
NM_000719.6 NM_001129827.1 NM_001129829.1 NM_001129830.2 NM_001129831.1 NM_001129832.1 NM_001129833.1 NM_001129834.1 NM_001129835.1 NM_001129836.1 NM_001129837.1 NM_001129838.1 NM_001129839.1 NM_001129840.1 NM_001129841.1 NM_001129842.1 NM_001129843.1 NM_001129844.1 NM_001129846.1 NM_001167623.1 NM_001167624.2 NM_001167625.1 NM_199460.3 XM_006719017.2 XM_011521020.2 XM_011521023.2 XM_017019926.1 XM_017019927.1 XM_017019928.1 XM_017019929.1 XM_017019930.1 XM_017019931.1 XM_017019932.1 XM_017019933.1 XM_017019934.1 XM_017019935.1 XM_017019936.1 XM_017019937.1 XM_017019938.1 XM_017019939.1 XM_017019940.1 XM_017019941.1 XM_017019942.1 XM_017019943.1 XM_017019944.1 XM_017019945.1 XM_017019946.1 XM_017019947.1 XM_017019948.1 XM_017019949.1 XM_017019950.1 XM_017019951.1 XM_017019952.1 XM_017019953.1 XM_017019954.1 XM_017019955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3459417 c.443C>T p.P148L Chr.12: 2120396-2120396 on Build GRCh38
3739563 c.408C>A p.A136A Chr.12: 2120361-2120361 on Build GRCh38
3739566 c.498C>A p.A166A Chr.12: 2120361-2120361 on Build GRCh38
3739564 c.408C>A p.A136A Chr.12: 2120361-2120361 on Build GRCh38
6617626 c.394C>T p.L132L Chr.12: 2120347-2120347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751407951 Chr.12: 2120449-2120449 on Build GRCh38
rs546786746 Chr.12: 2120379-2120379 on Build GRCh38
rs186692112 Chr.12: 2120467-2120467 on Build GRCh38
rs761891438 Chr.12: 2120300-2120300 on Build GRCh38
rs367638447 Chr.12: 2120412-2120412 on Build GRCh38
rs375212916 Chr.12: 2120296-2120296 on Build GRCh38
rs772557072 Chr.12: 2120373-2120373 on Build GRCh38
rs747719890 Chr.12: 2120360-2120360 on Build GRCh38
rs748949538 Chr.12: 2120372-2120372 on Build GRCh38
rs760538776 Chr.12: 2120297-2120297 on Build GRCh38
rs776532518 Chr.12: 2120409-2120409 on Build GRCh38
rs780660896 Chr.12: 2120350-2120350 on Build GRCh38
rs763228787 Chr.12: 2120438-2120438 on Build GRCh38
rs367763190 Chr.12: 2120316-2120316 on Build GRCh38
rs55792866 Chr.12: 2120310-2120310 on Build GRCh38
rs754865091 Chr.12: 2120453-2120453 on Build GRCh38
rs961457767 Chr.12: 2120437-2120437 on Build GRCh38
rs769308149 Chr.12: 2120397-2120397 on Build GRCh38
rs769295096 Chr.12: 2120361-2120361 on Build GRCh38
rs750120212 Chr.12: 2120306-2120306 on Build GRCh38
rs533097898 Chr.12: 2120367-2120367 on Build GRCh38
rs587780882 Chr.12: 2120328-2120328 on Build GRCh38
rs750645940 Chr.12: 2120416-2120416 on Build GRCh38
rs761958170 Chr.12: 2120411-2120411 on Build GRCh38
rs200282707 Chr.12: 2120302-2120302 on Build GRCh38
rs1019342847 Chr.12: 2120427-2120427 on Build GRCh38
rs761239778 Chr.12: 2120384-2120384 on Build GRCh38
rs371836468 Chr.12: 2120444-2120444 on Build GRCh38
rs754951873 Chr.12: 2120335-2120335 on Build GRCh38

View Full Product Details