Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161711307-161711532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRLA
Entrez Gene ID
84824
Gene Name
Fc receptor like A
Gene Aliases
FCRL, FCRL1, FCRLM1, FCRLX, FCRLb, FCRLc1, FCRLc2, FCRLd, FCRLe, FCRX, FREB
UniGene
Hs.266331
Gene Chromosome Location
Chr.1: 161706972-161714352 on Build GRCh38
Transcripts
NM_001184866.1 NM_001184867.1 NM_001184870.1 NM_001184871.1 NM_001184872.1 NM_001184873.1 NM_032738.3 XM_006711581.3 XM_011510064.1 XM_011510065.1 XM_011510066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5170465 c.393C>A p.N131K Chr.1: 161711350-161711350 on Build GRCh38
3965967 c.366C>T p.P122P Chr.1: 161711341-161711341 on Build GRCh38
1335695 c.564C>T p.V188V Chr.1: 161711470-161711470 on Build GRCh38
7368098 c.472G>A p.A158T Chr.1: 161711447-161711447 on Build GRCh38
2087386 c.361C>T p.P121S Chr.1: 161711336-161711336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753865073 Chr.1: 161711357-161711358 on Build GRCh38
rs766930615 Chr.1: 161711437-161711437 on Build GRCh38
rs889373513 Chr.1: 161711426-161711426 on Build GRCh38
rs745824269 Chr.1: 161711475-161711475 on Build GRCh38
rs752347588 Chr.1: 161711329-161711329 on Build GRCh38
rs199504307 Chr.1: 161711509-161711509 on Build GRCh38
rs756284231 Chr.1: 161711458-161711458 on Build GRCh38
rs761404270 Chr.1: 161711435-161711435 on Build GRCh38
rs745591681 Chr.1: 161711390-161711390 on Build GRCh38
rs764863112 Chr.1: 161711500-161711500 on Build GRCh38
rs775114285 Chr.1: 161711488-161711488 on Build GRCh38
rs756410172 Chr.1: 161711356-161711356 on Build GRCh38
rs781524210 Chr.1: 161711354-161711354 on Build GRCh38
rs760219794 Chr.1: 161711421-161711421 on Build GRCh38
rs969291336 Chr.1: 161711506-161711506 on Build GRCh38
rs778113827 Chr.1: 161711459-161711459 on Build GRCh38
rs144830049 Chr.1: 161711448-161711448 on Build GRCh38
rs537598966 Chr.1: 161711368-161711368 on Build GRCh38
rs1019080860 Chr.1: 161711434-161711434 on Build GRCh38
rs755400433 Chr.1: 161711353-161711353 on Build GRCh38
rs750548512 Chr.1: 161711389-161711389 on Build GRCh38
rs763877414 Chr.1: 161711422-161711422 on Build GRCh38
rs752143645 Chr.1: 161711438-161711438 on Build GRCh38
rs749268660 Chr.1: 161711464-161711464 on Build GRCh38
rs142089406 Chr.1: 161711369-161711369 on Build GRCh38
rs989137134 Chr.1: 161711471-161711471 on Build GRCh38
rs746882382 Chr.1: 161711489-161711489 on Build GRCh38
rs751918714 Chr.1: 161711351-161711351 on Build GRCh38
rs761370681 Chr.1: 161711497-161711497 on Build GRCh38
rs886162238 Chr.1: 161711513-161711513 on Build GRCh38
rs774673958 Chr.1: 161711454-161711454 on Build GRCh38
rs771880619 Chr.1: 161711406-161711406 on Build GRCh38
rs775156996 Chr.1: 161711407-161711407 on Build GRCh38
rs772043962 Chr.1: 161711476-161711476 on Build GRCh38
rs371004106 Chr.1: 161711342-161711342 on Build GRCh38
rs757332310 Chr.1: 161711466-161711466 on Build GRCh38
rs376754195 Chr.1: 161711341-161711341 on Build GRCh38
rs112766190 Chr.1: 161711334-161711334 on Build GRCh38
rs1003415898 Chr.1: 161711346-161711346 on Build GRCh38
rs138171030 Chr.1: 161711432-161711432 on Build GRCh38
rs201524104 Chr.1: 161711499-161711499 on Build GRCh38
rs768138226 Chr.1: 161711491-161711491 on Build GRCh38
rs776354718 Chr.1: 161711492-161711492 on Build GRCh38
rs564868949 Chr.1: 161711362-161711362 on Build GRCh38
rs199766155 Chr.1: 161711414-161711414 on Build GRCh38
rs376314134 Chr.1: 161711375-161711375 on Build GRCh38
rs753147888 Chr.1: 161711452-161711452 on Build GRCh38
rs778779192 Chr.1: 161711472-161711472 on Build GRCh38
rs763531432 Chr.1: 161711336-161711336 on Build GRCh38
rs760090571 Chr.1: 161711510-161711510 on Build GRCh38
rs144403707 Chr.1: 161711447-161711447 on Build GRCh38
rs767055541 Chr.1: 161711350-161711350 on Build GRCh38
rs1022174220 Chr.1: 161711419-161711419 on Build GRCh38

View Full Product Details