Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122577702-122577943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMBT1
Entrez Gene ID
1755
Gene Name
deleted in malignant brain tumors 1
Gene Aliases
GP340, SAG, muclin
UniGene
Hs.279611
Gene Chromosome Location
Chr.10: 122560665-122643736 on Build GRCh38
Transcripts
NM_001320644.1 NM_004406.2 NM_007329.2 NM_017579.2 XM_006717660.3 XM_006717665.3 XM_011539388.2 XM_011539389.2 XM_011539390.2 XM_011539391.2 XM_011539392.2 XM_011539393.2 XM_011539394.2 XM_011539395.2 XM_011539396.2 XM_011539398.2 XM_011539399.2 XM_011539400.2 XM_011539401.2 XM_011539402.2 XM_011539403.2 XM_011539405.2 XM_011539407.2 XM_011539408.2 XM_011539409.2 XM_011539410.2 XM_011539411.2 XM_011539413.2 XM_011539414.2 XM_011539415.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4012292 c.612C>A p.A204A Chr.10: 122577815-122577815 on Build GRCh38
7269651 c.610G>A p.A204T Chr.10: 122577813-122577813 on Build GRCh38
4012293 c.612C>A p.A204A Chr.10: 122577815-122577815 on Build GRCh38
7269652 c.610G>A p.A204T Chr.10: 122577813-122577813 on Build GRCh38
223424 c.637+1G>A p.? Chr.10: 122577841-122577841 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765242297 Chr.10: 122577819-122577819 on Build GRCh38
rs200051753 Chr.10: 122577809-122577809 on Build GRCh38
rs769996553 Chr.10: 122577841-122577841 on Build GRCh38
rs763819834 Chr.10: 122577846-122577847 on Build GRCh38
rs768083356 Chr.10: 122577791-122577791 on Build GRCh38
rs747474701 Chr.10: 122577872-122577872 on Build GRCh38
rs776747053 Chr.10: 122577812-122577812 on Build GRCh38
rs950111096 Chr.10: 122577757-122577757 on Build GRCh38
rs200245641 Chr.10: 122577829-122577829 on Build GRCh38
rs184297664 Chr.10: 122577866-122577866 on Build GRCh38
rs57544182 Chr.10: 122577864-122577864 on Build GRCh38
rs755888132 Chr.10: 122577782-122577782 on Build GRCh38
rs202194426 Chr.10: 122577849-122577849 on Build GRCh38
rs771440468 Chr.10: 122577879-122577879 on Build GRCh38
rs753458105 Chr.10: 122577856-122577856 on Build GRCh38
rs767498588 Chr.10: 122577836-122577836 on Build GRCh38
rs779730149 Chr.10: 122577786-122577786 on Build GRCh38
rs748940652 Chr.10: 122577789-122577789 on Build GRCh38
rs778438837 Chr.10: 122577796-122577796 on Build GRCh38
rs759752756 Chr.10: 122577814-122577814 on Build GRCh38
rs552209441 Chr.10: 122577868-122577868 on Build GRCh38
rs761182515 Chr.10: 122577808-122577808 on Build GRCh38
rs540523953 Chr.10: 122577735-122577735 on Build GRCh38
rs748179866 Chr.10: 122577721-122577722 on Build GRCh38
rs746079835 Chr.10: 122577888-122577888 on Build GRCh38
rs778293638 Chr.10: 122577863-122577863 on Build GRCh38
rs530511573 Chr.10: 122577806-122577806 on Build GRCh38
rs752753413 Chr.10: 122577821-122577821 on Build GRCh38
rs754602907 Chr.10: 122577858-122577858 on Build GRCh38
rs1024981607 Chr.10: 122577759-122577759 on Build GRCh38
rs936087881 Chr.10: 122577723-122577723 on Build GRCh38
rs1055700848 Chr.10: 122577724-122577724 on Build GRCh38
rs1005117438 Chr.10: 122577793-122577793 on Build GRCh38
rs79660171 Chr.10: 122577913-122577913 on Build GRCh38
rs772644685 Chr.10: 122577798-122577798 on Build GRCh38
rs779785466 Chr.10: 122577855-122577855 on Build GRCh38
rs577870797 Chr.10: 122577726-122577726 on Build GRCh38
rs551439423 Chr.10: 122577884-122577884 on Build GRCh38
rs750284644 Chr.10: 122577781-122577781 on Build GRCh38
rs748616774 Chr.10: 122577797-122577797 on Build GRCh38
rs761683602 Chr.10: 122577867-122577867 on Build GRCh38

View Full Product Details