Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89197883-89198126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRR1
Entrez Gene ID
2569
Gene Name
gamma-aminobutyric acid type A receptor rho1 subunit
Gene Aliases
-
UniGene
Hs.99927
Gene Chromosome Location
Chr.6: 89177504-89231288 on Build GRCh38
Transcripts
NM_001256703.1 NM_001256704.1 NM_001267582.1 NM_002042.4 XM_017010689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3178077 c.535C>T p.R179W Chr.6: 89198057-89198057 on Build GRCh38
6321254 c.510C>A p.D170E Chr.6: 89198082-89198082 on Build GRCh38
6297604 c.518C>A p.T173K Chr.6: 89198074-89198074 on Build GRCh38
4686181 c.481G>A p.V161M Chr.6: 89198111-89198111 on Build GRCh38
6668728 c.507C>T p.H169H Chr.6: 89198085-89198085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996918024 Chr.6: 89198031-89198031 on Build GRCh38
rs759235678 Chr.6: 89197986-89197986 on Build GRCh38
rs989949555 Chr.6: 89198083-89198083 on Build GRCh38
rs757293852 Chr.6: 89197976-89197976 on Build GRCh38
rs781156704 Chr.6: 89198078-89198078 on Build GRCh38
rs774850815 Chr.6: 89198009-89198009 on Build GRCh38
rs201013212 Chr.6: 89198057-89198057 on Build GRCh38
rs749220392 Chr.6: 89198079-89198079 on Build GRCh38
rs760176036 Chr.6: 89198013-89198013 on Build GRCh38
rs566948105 Chr.6: 89197949-89197949 on Build GRCh38
rs781712512 Chr.6: 89198007-89198007 on Build GRCh38
rs778850287 Chr.6: 89198089-89198089 on Build GRCh38
rs964401673 Chr.6: 89198027-89198027 on Build GRCh38
rs34218666 Chr.6: 89198067-89198067 on Build GRCh38
rs763517347 Chr.6: 89198025-89198025 on Build GRCh38
rs200777807 Chr.6: 89198090-89198090 on Build GRCh38
rs758457192 Chr.6: 89197949-89197949 on Build GRCh38
rs771920686 Chr.6: 89197996-89197996 on Build GRCh38
rs754331590 Chr.6: 89198029-89198029 on Build GRCh38
rs771620125 Chr.6: 89198017-89198017 on Build GRCh38
rs751732693 Chr.6: 89198066-89198066 on Build GRCh38
rs1019585628 Chr.6: 89198020-89198020 on Build GRCh38
rs77084777 Chr.6: 89197947-89197947 on Build GRCh38
rs775112992 Chr.6: 89198040-89198040 on Build GRCh38
rs780261898 Chr.6: 89198062-89198062 on Build GRCh38
rs1047294081 Chr.6: 89197955-89197955 on Build GRCh38
rs370358388 Chr.6: 89197980-89197980 on Build GRCh38
rs745715885 Chr.6: 89198091-89198091 on Build GRCh38
rs771033888 Chr.6: 89198082-89198082 on Build GRCh38
rs762356649 Chr.6: 89198042-89198042 on Build GRCh38
rs866664802 Chr.6: 89198016-89198016 on Build GRCh38
rs534023247 Chr.6: 89198049-89198049 on Build GRCh38
rs934702050 Chr.6: 89197954-89197954 on Build GRCh38
rs376706946 Chr.6: 89197977-89197977 on Build GRCh38
rs185436805 Chr.6: 89198099-89198099 on Build GRCh38
rs201812835 Chr.6: 89198098-89198098 on Build GRCh38
rs750851828 Chr.6: 89198056-89198056 on Build GRCh38

View Full Product Details