Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65198349-65198588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CILP
Entrez Gene ID
8483
Gene Name
cartilage intermediate layer protein
Gene Aliases
CILP-1, HsT18872
UniGene
Hs.442180
Gene Chromosome Location
Chr.15: 65195999-65211608 on Build GRCh38
Transcripts
NM_003613.3 XM_017022678.1 XM_017022679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3503157 c.1878C>T p.F626F Chr.15: 65198408-65198408 on Build GRCh38
3503156 c.1887C>T p.P629P Chr.15: 65198399-65198399 on Build GRCh38
4578427 c.1922A>G p.D641G Chr.15: 65198364-65198364 on Build GRCh38
3503159 c.1853G>A p.G618E Chr.15: 65198433-65198433 on Build GRCh38
5717814 c.1879C>T p.L627L Chr.15: 65198407-65198407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757859071 Chr.15: 65198517-65198517 on Build GRCh38
rs887208515 Chr.15: 65198442-65198442 on Build GRCh38
rs749708471 Chr.15: 65198533-65198533 on Build GRCh38
rs767788040 Chr.15: 65198521-65198521 on Build GRCh38
rs745335391 Chr.15: 65198510-65198510 on Build GRCh38
rs376138832 Chr.15: 65198521-65198521 on Build GRCh38
rs2679118 Chr.15: 65198563-65198563 on Build GRCh38
rs773778153 Chr.15: 65198435-65198435 on Build GRCh38
rs942723895 Chr.15: 65198525-65198525 on Build GRCh38
rs199711765 Chr.15: 65198397-65198397 on Build GRCh38
rs761417111 Chr.15: 65198473-65198473 on Build GRCh38
rs748486808 Chr.15: 65198419-65198419 on Build GRCh38
rs778461184 Chr.15: 65198505-65198505 on Build GRCh38
rs774455344 Chr.15: 65198558-65198558 on Build GRCh38
rs200703143 Chr.15: 65198389-65198389 on Build GRCh38
rs767434294 Chr.15: 65198483-65198483 on Build GRCh38
rs760004371 Chr.15: 65198509-65198509 on Build GRCh38
rs374945156 Chr.15: 65198526-65198526 on Build GRCh38
rs756962779 Chr.15: 65198388-65198388 on Build GRCh38
rs766754083 Chr.15: 65198447-65198447 on Build GRCh38
rs752585496 Chr.15: 65198489-65198489 on Build GRCh38
rs564148044 Chr.15: 65198418-65198418 on Build GRCh38
rs911205184 Chr.15: 65198504-65198504 on Build GRCh38
rs770479877 Chr.15: 65198522-65198522 on Build GRCh38
rs751867966 Chr.15: 65198462-65198462 on Build GRCh38
rs201651652 Chr.15: 65198386-65198386 on Build GRCh38
rs753684058 Chr.15: 65198498-65198498 on Build GRCh38
rs756113113 Chr.15: 65198494-65198494 on Build GRCh38
rs763251583 Chr.15: 65198436-65198436 on Build GRCh38
rs774454242 Chr.15: 65198459-65198459 on Build GRCh38
rs759598002 Chr.15: 65198562-65198562 on Build GRCh38
rs143772526 Chr.15: 65198497-65198497 on Build GRCh38
rs764768979 Chr.15: 65198555-65198555 on Build GRCh38
rs866356188 Chr.15: 65198532-65198532 on Build GRCh38
rs776406887 Chr.15: 65198401-65198401 on Build GRCh38
rs771234050 Chr.15: 65198543-65198543 on Build GRCh38
rs777025663 Chr.15: 65198412-65198412 on Build GRCh38
rs770285469 Chr.15: 65198426-65198426 on Build GRCh38
rs530704700 Chr.15: 65198398-65198398 on Build GRCh38

View Full Product Details