Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85371017-85371290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMOD3
Entrez Gene ID
84173
Gene Name
ELMO domain containing 3
Gene Aliases
DFNB88, LST3, RBED1, RBM29
UniGene
Hs.269990
Gene Chromosome Location
Chr.2: 85354394-85391752 on Build GRCh38
Transcripts
NM_001135021.1 NM_001135022.1 NM_001135023.1 NM_032213.4 XM_005264596.3 XM_005264597.3 XM_005264598.3 XM_017005078.1 XM_017005079.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1592436 c.400G>A p.A134T Chr.2: 85371125-85371125 on Build GRCh38
4996261 c.448C>T p.R150W Chr.2: 85371173-85371173 on Build GRCh38
3426728 c.420C>T p.F140F Chr.2: 85371145-85371145 on Build GRCh38
723029 c.399C>A p.A133A Chr.2: 85371124-85371124 on Build GRCh38
4880690 c.484+4A>G p.? Chr.2: 85371213-85371213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201692835 Chr.2: 85371201-85371201 on Build GRCh38
rs373275031 Chr.2: 85371222-85371222 on Build GRCh38
rs143960314 Chr.2: 85371152-85371152 on Build GRCh38
rs752397334 Chr.2: 85371271-85371271 on Build GRCh38
rs569639102 Chr.2: 85371219-85371219 on Build GRCh38
rs201052944 Chr.2: 85371131-85371131 on Build GRCh38
rs766073287 Chr.2: 85371167-85371167 on Build GRCh38
rs529093966 Chr.2: 85371132-85371132 on Build GRCh38
rs1040129255 Chr.2: 85371165-85371165 on Build GRCh38
rs767176022 Chr.2: 85371145-85371145 on Build GRCh38
rs545039229 Chr.2: 85371093-85371093 on Build GRCh38
rs753581498 Chr.2: 85371168-85371168 on Build GRCh38
rs760198766 Chr.2: 85371225-85371225 on Build GRCh38
rs752707208 Chr.2: 85371196-85371196 on Build GRCh38
rs775846168 Chr.2: 85371207-85371207 on Build GRCh38
rs771060620 Chr.2: 85371113-85371113 on Build GRCh38
rs756219282 Chr.2: 85371265-85371266 on Build GRCh38
rs371254266 Chr.2: 85371125-85371125 on Build GRCh38
rs755795191 Chr.2: 85371084-85371084 on Build GRCh38
rs969115966 Chr.2: 85371052-85371052 on Build GRCh38
rs763308374 Chr.2: 85371128-85371128 on Build GRCh38
rs767121154 Chr.2: 85371058-85371058 on Build GRCh38
rs369816241 Chr.2: 85371186-85371186 on Build GRCh38
rs765982657 Chr.2: 85371226-85371226 on Build GRCh38
rs948696959 Chr.2: 85371150-85371150 on Build GRCh38
rs1017070065 Chr.2: 85371257-85371257 on Build GRCh38
rs145555897 Chr.2: 85371122-85371122 on Build GRCh38
rs765124410 Chr.2: 85371267-85371267 on Build GRCh38
rs753586335 Chr.2: 85371265-85371265 on Build GRCh38
rs142656005 Chr.2: 85371173-85371173 on Build GRCh38
rs1025667495 Chr.2: 85371218-85371218 on Build GRCh38
rs202151775 Chr.2: 85371220-85371220 on Build GRCh38
rs375552296 Chr.2: 85371223-85371223 on Build GRCh38
rs756855018 Chr.2: 85371057-85371057 on Build GRCh38
rs769112797 Chr.2: 85371212-85371212 on Build GRCh38
rs745902219 Chr.2: 85371187-85371187 on Build GRCh38
rs557345712 Chr.2: 85371101-85371101 on Build GRCh38
rs200710196 Chr.2: 85371210-85371210 on Build GRCh38
rs749063389 Chr.2: 85371097-85371097 on Build GRCh38
rs199660149 Chr.2: 85371148-85371148 on Build GRCh38
rs748169877 Chr.2: 85371174-85371174 on Build GRCh38
rs753998682 Chr.2: 85371215-85371215 on Build GRCh38
rs867831354 Chr.2: 85371041-85371041 on Build GRCh38
rs750149957 Chr.2: 85371064-85371064 on Build GRCh38
rs562825417 Chr.2: 85371107-85371107 on Build GRCh38
rs927627619 Chr.2: 85371103-85371103 on Build GRCh38
rs114220580 Chr.2: 85371266-85371266 on Build GRCh38
rs762203882 Chr.2: 85371139-85371139 on Build GRCh38
rs889037351 Chr.2: 85371133-85371133 on Build GRCh38
rs112857214 Chr.2: 85371121-85371121 on Build GRCh38
rs767794151 Chr.2: 85371157-85371157 on Build GRCh38
rs757409054 Chr.2: 85371264-85371264 on Build GRCh38
rs769966776 Chr.2: 85371124-85371124 on Build GRCh38
rs575600796 Chr.2: 85371108-85371108 on Build GRCh38
rs758305442 Chr.2: 85371182-85371182 on Build GRCh38

View Full Product Details