Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68684881-68685122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAL
Entrez Gene ID
51083
Gene Name
galanin and GMAP prepropeptide
Gene Aliases
ETL8, GAL-GMAP, GALN, GLNN, GMAP
UniGene
Hs.278959
Gene Chromosome Location
Chr.11: 68684475-68691175 on Build GRCh38
Transcripts
NM_015973.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4439608 c.42C>T p.L14L Chr.11: 68684965-68684965 on Build GRCh38
7369121 c.65C>A p.A22E Chr.11: 68684988-68684988 on Build GRCh38
6401309 c.16G>A p.A6T Chr.11: 68684939-68684939 on Build GRCh38
6380583 c.7C>T p.R3* Chr.11: 68684930-68684930 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746008445 Chr.11: 68684978-68684978 on Build GRCh38
rs758825923 Chr.11: 68685042-68685042 on Build GRCh38
rs747717547 Chr.11: 68685004-68685004 on Build GRCh38
rs908596910 Chr.11: 68684950-68684950 on Build GRCh38
rs766211433 Chr.11: 68684951-68684951 on Build GRCh38
rs530826193 Chr.11: 68684988-68684988 on Build GRCh38
rs958349705 Chr.11: 68685054-68685054 on Build GRCh38
rs542684427 Chr.11: 68684989-68684989 on Build GRCh38
rs760189438 Chr.11: 68685029-68685029 on Build GRCh38
rs539085763 Chr.11: 68684932-68684932 on Build GRCh38
rs749996027 Chr.11: 68684936-68684936 on Build GRCh38
rs143156739 Chr.11: 68684934-68684934 on Build GRCh38
rs755026539 Chr.11: 68684967-68684967 on Build GRCh38
rs764776196 Chr.11: 68685022-68685024 on Build GRCh38
rs991736017 Chr.11: 68684917-68684917 on Build GRCh38
rs772924321 Chr.11: 68685027-68685027 on Build GRCh38
rs996764651 Chr.11: 68685094-68685094 on Build GRCh38
rs185026810 Chr.11: 68684902-68684902 on Build GRCh38
rs956379887 Chr.11: 68684960-68684960 on Build GRCh38
rs939120221 Chr.11: 68684994-68684994 on Build GRCh38
rs578193461 Chr.11: 68684930-68684930 on Build GRCh38
rs373559015 Chr.11: 68685032-68685032 on Build GRCh38
rs763391717 Chr.11: 68684923-68684923 on Build GRCh38
rs561023780 Chr.11: 68685031-68685031 on Build GRCh38
rs770012087 Chr.11: 68684912-68684912 on Build GRCh38
rs17852742 Chr.11: 68684947-68684947 on Build GRCh38
rs771255713 Chr.11: 68684901-68684901 on Build GRCh38
rs1035394004 Chr.11: 68685053-68685053 on Build GRCh38
rs760551954 Chr.11: 68684939-68684939 on Build GRCh38
rs751705346 Chr.11: 68684968-68684968 on Build GRCh38
rs774699437 Chr.11: 68685003-68685003 on Build GRCh38
rs1011788651 Chr.11: 68685030-68685030 on Build GRCh38
rs746262791 Chr.11: 68684904-68684904 on Build GRCh38
rs753843211 Chr.11: 68684966-68684966 on Build GRCh38
rs747050822 Chr.11: 68684900-68684900 on Build GRCh38
rs990258729 Chr.11: 68684963-68684963 on Build GRCh38
rs759378653 Chr.11: 68685049-68685049 on Build GRCh38
rs931437075 Chr.11: 68684931-68684931 on Build GRCh38
rs775978483 Chr.11: 68684921-68684921 on Build GRCh38
rs183235150 Chr.11: 68685088-68685088 on Build GRCh38
rs749648649 Chr.11: 68684992-68684992 on Build GRCh38
rs765284289 Chr.11: 68685055-68685055 on Build GRCh38
rs34725707 Chr.11: 68684970-68684970 on Build GRCh38
rs201644659 Chr.11: 68685000-68685000 on Build GRCh38
rs1038198795 Chr.11: 68685050-68685050 on Build GRCh38
rs781505935 Chr.11: 68684976-68684976 on Build GRCh38
rs761299568 Chr.11: 68684994-68684995 on Build GRCh38
rs200711676 Chr.11: 68685039-68685039 on Build GRCh38
rs528596322 Chr.11: 68685051-68685051 on Build GRCh38
rs146709951 Chr.11: 68684941-68684941 on Build GRCh38

View Full Product Details