Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44751190-44751439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL3
Entrez Gene ID
602
Gene Name
B-cell CLL/lymphoma 3
Gene Aliases
BCL4, D19S37
UniGene
Hs.31210
Gene Chromosome Location
Chr.19: 44742621-44760044 on Build GRCh38
Transcripts
NM_005178.4 XM_011527197.2 XM_011527198.2 XM_011527200.2 XM_017027109.1 XM_017027110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3720969 c.284G>A p.R95Q Chr.19: 44751254-44751254 on Build GRCh38
3535741 c.346C>T p.P116S Chr.19: 44751316-44751316 on Build GRCh38
3103325 c.299C>T p.P100L Chr.19: 44751269-44751269 on Build GRCh38
1184593 c.344G>A p.C115Y Chr.19: 44751314-44751314 on Build GRCh38
5476082 c.402C>T p.D134D Chr.19: 44751372-44751372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899315490 Chr.19: 44751254-44751254 on Build GRCh38
rs1036968606 Chr.19: 44751252-44751252 on Build GRCh38
rs781291617 Chr.19: 44751270-44751270 on Build GRCh38
rs745707405 Chr.19: 44751359-44751359 on Build GRCh38
rs755034202 Chr.19: 44751269-44751269 on Build GRCh38
rs753460174 Chr.19: 44751218-44751218 on Build GRCh38
rs760072914 Chr.19: 44751211-44751211 on Build GRCh38
rs1011893746 Chr.19: 44751298-44751298 on Build GRCh38
rs763277716 Chr.19: 44751320-44751320 on Build GRCh38
rs747158597 Chr.19: 44751256-44751256 on Build GRCh38
rs775687244 Chr.19: 44751392-44751392 on Build GRCh38
rs776069320 Chr.19: 44751380-44751380 on Build GRCh38
rs35777633 Chr.19: 44751258-44751259 on Build GRCh38
rs760615560 Chr.19: 44751305-44751305 on Build GRCh38
rs139775439 Chr.19: 44751366-44751366 on Build GRCh38
rs768583508 Chr.19: 44751373-44751373 on Build GRCh38
rs765593954 Chr.19: 44751230-44751230 on Build GRCh38
rs371024213 Chr.19: 44751304-44751304 on Build GRCh38
rs774348285 Chr.19: 44751286-44751286 on Build GRCh38
rs140257795 Chr.19: 44751346-44751346 on Build GRCh38
rs898149672 Chr.19: 44751219-44751219 on Build GRCh38
rs376985588 Chr.19: 44751409-44751409 on Build GRCh38
rs745656312 Chr.19: 44751297-44751297 on Build GRCh38
rs758969241 Chr.19: 44751250-44751250 on Build GRCh38
rs901965741 Chr.19: 44751325-44751325 on Build GRCh38
rs775268394 Chr.19: 44751301-44751301 on Build GRCh38
rs766714413 Chr.19: 44751327-44751327 on Build GRCh38
rs369493245 Chr.19: 44751249-44751249 on Build GRCh38
rs768087111 Chr.19: 44751307-44751307 on Build GRCh38
rs755444152 Chr.19: 44751333-44751333 on Build GRCh38
rs772870500 Chr.19: 44751398-44751398 on Build GRCh38
rs201526615 Chr.19: 44751227-44751227 on Build GRCh38
rs764862709 Chr.19: 44751309-44751309 on Build GRCh38
rs527504186 Chr.19: 44751369-44751369 on Build GRCh38
rs756866502 Chr.19: 44751224-44751224 on Build GRCh38
rs780503593 Chr.19: 44751253-44751253 on Build GRCh38
rs373600361 Chr.19: 44751285-44751285 on Build GRCh38
rs752037614 Chr.19: 44751331-44751331 on Build GRCh38
rs150348108 Chr.19: 44751372-44751372 on Build GRCh38
rs751891657 Chr.19: 44751414-44751414 on Build GRCh38
rs748322046 Chr.19: 44751276-44751276 on Build GRCh38
rs756473223 Chr.19: 44751356-44751356 on Build GRCh38
rs761279156 Chr.19: 44751393-44751393 on Build GRCh38
rs750824997 Chr.19: 44751319-44751319 on Build GRCh38
rs147904593 Chr.19: 44751345-44751345 on Build GRCh38
rs778110510 Chr.19: 44751358-44751358 on Build GRCh38
rs552485571 Chr.19: 44751308-44751308 on Build GRCh38
rs763860988 Chr.19: 44751212-44751212 on Build GRCh38
rs774635426 Chr.19: 44751392-44751392 on Build GRCh38
rs376501290 Chr.19: 44751300-44751300 on Build GRCh38
rs762551283 Chr.19: 44751404-44751404 on Build GRCh38

View Full Product Details