Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64088786-64089011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM96A
Entrez Gene ID
84191
Gene Name
family with sequence similarity 96 member A
Gene Aliases
-
UniGene
Hs.439548
Gene Chromosome Location
Chr.15: 64072559-64094008 on Build GRCh38
Transcripts
NM_001014812.2 NM_001289108.1 NM_032231.5 NR_110310.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
138971 c.151G>C p.E51Q Chr.15: 64088825-64088825 on Build GRCh38
470948 c.183G>T p.V61V Chr.15: 64088793-64088793 on Build GRCh38
4683309 c.188C>T p.S63L Chr.15: 64088788-64088788 on Build GRCh38
963902 c.158C>T p.P53L Chr.15: 64088818-64088818 on Build GRCh38
6446030 c.125-1G>A p.? Chr.15: 64088852-64088852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043749489 Chr.15: 64088946-64088946 on Build GRCh38
rs780270751 Chr.15: 64088829-64088829 on Build GRCh38
rs978573240 Chr.15: 64088903-64088903 on Build GRCh38
rs373873752 Chr.15: 64088890-64088890 on Build GRCh38
rs924754505 Chr.15: 64088909-64088909 on Build GRCh38
rs993417146 Chr.15: 64088935-64088935 on Build GRCh38
rs965303506 Chr.15: 64088878-64088878 on Build GRCh38
rs934873425 Chr.15: 64088923-64088923 on Build GRCh38
rs759190098 Chr.15: 64088882-64088882 on Build GRCh38
rs751060921 Chr.15: 64088881-64088881 on Build GRCh38
rs946664267 Chr.15: 64088894-64088894 on Build GRCh38
rs772329697 Chr.15: 64088816-64088816 on Build GRCh38
rs773061022 Chr.15: 64088854-64088854 on Build GRCh38
rs769800607 Chr.15: 64088853-64088853 on Build GRCh38
rs747303027 Chr.15: 64088833-64088833 on Build GRCh38
rs147426625 Chr.15: 64088834-64088834 on Build GRCh38
rs754264351 Chr.15: 64088895-64088895 on Build GRCh38
rs754066400 Chr.15: 64088870-64088871 on Build GRCh38
rs764407417 Chr.15: 64088849-64088853 on Build GRCh38
rs776499715 Chr.15: 64088837-64088837 on Build GRCh38
rs140765077 Chr.15: 64088960-64088960 on Build GRCh38
rs16947748 Chr.15: 64088949-64088949 on Build GRCh38
rs779252477 Chr.15: 64088809-64088809 on Build GRCh38
rs199903062 Chr.15: 64088891-64088891 on Build GRCh38
rs142829471 Chr.15: 64088813-64088813 on Build GRCh38
rs761466140 Chr.15: 64088839-64088839 on Build GRCh38
rs762923165 Chr.15: 64088868-64088868 on Build GRCh38

View Full Product Details