Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36014082-36014339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH6
Entrez Gene ID
84964
Gene Name
alkB homolog 6
Gene Aliases
ABH6
UniGene
Hs.71941
Gene Chromosome Location
Chr.19: 36009116-36014243 on Build GRCh38
Transcripts
NM_001297701.1 NM_032878.3 NM_198867.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7114059 c.12_15delGGGG p.G5fs*4 Chr.19: 36014219-36014222 on Build GRCh38
3959962 c.40G>T p.G14* Chr.19: 36014194-36014194 on Build GRCh38
3532860 c.15G>A p.G5G Chr.19: 36014219-36014219 on Build GRCh38
6890772 c.19G>A p.G7R Chr.19: 36014215-36014215 on Build GRCh38
1177531 c.10A>G p.R4G Chr.19: 36014224-36014224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376400772 Chr.19: 36014261-36014261 on Build GRCh38
rs745476490 Chr.19: 36014212-36014212 on Build GRCh38
rs766540678 Chr.19: 36014149-36014149 on Build GRCh38
rs371938864 Chr.19: 36014195-36014195 on Build GRCh38
rs934708439 Chr.19: 36014312-36014312 on Build GRCh38
rs1053195809 Chr.19: 36014317-36014317 on Build GRCh38
rs756080086 Chr.19: 36014148-36014148 on Build GRCh38
rs759710185 Chr.19: 36014224-36014224 on Build GRCh38
rs374100269 Chr.19: 36014151-36014151 on Build GRCh38
rs765700430 Chr.19: 36014248-36014251 on Build GRCh38
rs759559792 Chr.19: 36014242-36014242 on Build GRCh38
rs201830479 Chr.19: 36014224-36014224 on Build GRCh38
rs200176352 Chr.19: 36014250-36014250 on Build GRCh38
rs755268349 Chr.19: 36014153-36014153 on Build GRCh38
rs753915807 Chr.19: 36014241-36014241 on Build GRCh38
rs758283773 Chr.19: 36014187-36014187 on Build GRCh38
rs1019050932 Chr.19: 36014101-36014101 on Build GRCh38
rs201016149 Chr.19: 36014191-36014191 on Build GRCh38
rs745333330 Chr.19: 36014258-36014258 on Build GRCh38
rs775144215 Chr.19: 36014218-36014218 on Build GRCh38
rs754108981 Chr.19: 36014282-36014295 on Build GRCh38
rs985062549 Chr.19: 36014220-36014220 on Build GRCh38
rs766341384 Chr.19: 36014233-36014233 on Build GRCh38
rs771247637 Chr.19: 36014133-36014134 on Build GRCh38
rs1020687404 Chr.19: 36014165-36014165 on Build GRCh38
rs765501369 Chr.19: 36014248-36014248 on Build GRCh38
rs761293956 Chr.19: 36014232-36014232 on Build GRCh38
rs749006513 Chr.19: 36014264-36014264 on Build GRCh38
rs757256190 Chr.19: 36014255-36014255 on Build GRCh38
rs529169250 Chr.19: 36014260-36014260 on Build GRCh38
rs773703929 Chr.19: 36014229-36014229 on Build GRCh38
rs199923401 Chr.19: 36014190-36014190 on Build GRCh38
rs367997454 Chr.19: 36014194-36014194 on Build GRCh38
rs772405049 Chr.19: 36014279-36014279 on Build GRCh38
rs773606241 Chr.19: 36014280-36014280 on Build GRCh38
rs926167971 Chr.19: 36014240-36014240 on Build GRCh38
rs760978419 Chr.19: 36014282-36014282 on Build GRCh38
rs867808327 Chr.19: 36014236-36014236 on Build GRCh38
rs868171706 Chr.19: 36014251-36014251 on Build GRCh38
rs777790588 Chr.19: 36014188-36014188 on Build GRCh38
rs779615310 Chr.19: 36014263-36014263 on Build GRCh38
rs965756958 Chr.19: 36014135-36014135 on Build GRCh38
rs370830905 Chr.19: 36014284-36014284 on Build GRCh38
rs950060267 Chr.19: 36014277-36014277 on Build GRCh38
rs776559290 Chr.19: 36014157-36014157 on Build GRCh38
rs762105505 Chr.19: 36014219-36014219 on Build GRCh38
rs373479361 Chr.19: 36014252-36014252 on Build GRCh38
rs779310233 Chr.19: 36014175-36014175 on Build GRCh38
rs747905484 Chr.19: 36014181-36014181 on Build GRCh38
rs1026081763 Chr.19: 36014186-36014186 on Build GRCh38
rs376338501 Chr.19: 36014134-36014134 on Build GRCh38

View Full Product Details