Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3869684-3869909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DFFB
Entrez Gene ID
1677
Gene Name
DNA fragmentation factor subunit beta
Gene Aliases
CAD, CPAN, DFF-40, DFF2, DFF40
UniGene
Hs.133089
Gene Chromosome Location
Chr.1: 3857143-3885429 on Build GRCh38
Transcripts
NM_001282669.1 NM_001320132.1 NM_001320136.1 NM_004402.3 NR_104222.1 NR_135150.1 NR_135151.1 NR_135152.1 XM_017000498.1 XM_017000499.1 XM_017000500.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5669887 c.681+1G>A p.? Chr.1: 3869776-3869776 on Build GRCh38
3489193 c.672C>T p.F224F Chr.1: 3869766-3869766 on Build GRCh38
5846573 c.663A>C p.E221D Chr.1: 3869757-3869757 on Build GRCh38
127795 c.624C>T p.D208D Chr.1: 3869718-3869718 on Build GRCh38
6376028 c.667T>C p.W223R Chr.1: 3869761-3869761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779762912 Chr.1: 3869823-3869823 on Build GRCh38
rs780830672 Chr.1: 3869806-3869806 on Build GRCh38
rs758891041 Chr.1: 3869774-3869774 on Build GRCh38
rs964282268 Chr.1: 3869832-3869832 on Build GRCh38
rs974350930 Chr.1: 3869836-3869836 on Build GRCh38
rs772054653 Chr.1: 3869734-3869734 on Build GRCh38
rs751120520 Chr.1: 3869796-3869796 on Build GRCh38
rs975856651 Chr.1: 3869839-3869839 on Build GRCh38
rs753157909 Chr.1: 3869773-3869773 on Build GRCh38
rs529376269 Chr.1: 3869853-3869853 on Build GRCh38
rs763225181 Chr.1: 3869753-3869753 on Build GRCh38
rs543144329 Chr.1: 3869754-3869754 on Build GRCh38
rs553038205 Chr.1: 3869766-3869766 on Build GRCh38
rs751839191 Chr.1: 3869888-3869888 on Build GRCh38
rs368455033 Chr.1: 3869809-3869809 on Build GRCh38
rs10909811 Chr.1: 3869846-3869846 on Build GRCh38
rs374397595 Chr.1: 3869735-3869735 on Build GRCh38
rs983192953 Chr.1: 3869885-3869885 on Build GRCh38
rs766759983 Chr.1: 3869740-3869740 on Build GRCh38
rs573164898 Chr.1: 3869779-3869779 on Build GRCh38
rs371006504 Chr.1: 3869780-3869780 on Build GRCh38
rs530739719 Chr.1: 3869814-3869814 on Build GRCh38
rs930322980 Chr.1: 3869867-3869867 on Build GRCh38
rs944557266 Chr.1: 3869849-3869849 on Build GRCh38
rs573651889 Chr.1: 3869733-3869733 on Build GRCh38
rs921579586 Chr.1: 3869840-3869840 on Build GRCh38
rs374015758 Chr.1: 3869808-3869808 on Build GRCh38
rs986970413 Chr.1: 3869810-3869810 on Build GRCh38
rs780789759 Chr.1: 3869715-3869715 on Build GRCh38
rs923690061 Chr.1: 3869807-3869807 on Build GRCh38
rs542622663 Chr.1: 3869738-3869738 on Build GRCh38
rs202084499 Chr.1: 3869806-3869806 on Build GRCh38
rs1020802812 Chr.1: 3869794-3869794 on Build GRCh38
rs745710168 Chr.1: 3869716-3869716 on Build GRCh38
rs543520878 Chr.1: 3869831-3869831 on Build GRCh38
rs374612226 Chr.1: 3869787-3869787 on Build GRCh38
rs931673470 Chr.1: 3869841-3869841 on Build GRCh38
rs765633248 Chr.1: 3869768-3869768 on Build GRCh38
rs775469843 Chr.1: 3869719-3869719 on Build GRCh38

View Full Product Details