Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 26914449-26914678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASUN
Entrez Gene ID
55726
Gene Name
asunder, spermatogenesis regulator
Gene Aliases
C12orf11, GCT1, Mat89Bb, NET48, SPATA30
UniGene
Hs.505077
Gene Chromosome Location
Chr.12: 26905171-26938326 on Build GRCh38
Transcripts
NM_018164.2 XM_005253422.3 XM_011520750.2 XM_011520751.2 XM_011520752.2 XM_011520753.2 XM_017019632.1 XM_017019633.1 XM_017019634.1 XM_017019635.1 XM_017019636.1 XM_017019637.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6136450 c.1275C>G p.N425K Chr.12: 26914552-26914552 on Build GRCh38
6894623 c.1324G>C p.E442Q Chr.12: 26914503-26914503 on Build GRCh38
6136449 c.1249G>T p.D417Y Chr.12: 26914578-26914578 on Build GRCh38
6496019 c.1301G>A p.R434K Chr.12: 26914526-26914526 on Build GRCh38
938315 c.1311C>T p.I437I Chr.12: 26914516-26914516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765019515 Chr.12: 26914539-26914539 on Build GRCh38
rs769334657 Chr.12: 26914622-26914622 on Build GRCh38
rs763577127 Chr.12: 26914553-26914553 on Build GRCh38
rs776278300 Chr.12: 26914514-26914514 on Build GRCh38
rs774877634 Chr.12: 26914585-26914585 on Build GRCh38
rs767847733 Chr.12: 26914502-26914502 on Build GRCh38
rs376533817 Chr.12: 26914487-26914487 on Build GRCh38
rs750693368 Chr.12: 26914591-26914591 on Build GRCh38
rs751113804 Chr.12: 26914558-26914558 on Build GRCh38
rs1042106632 Chr.12: 26914506-26914506 on Build GRCh38
rs898343439 Chr.12: 26914646-26914646 on Build GRCh38
rs780952467 Chr.12: 26914589-26914589 on Build GRCh38
rs749835201 Chr.12: 26914619-26914619 on Build GRCh38
rs752666171 Chr.12: 26914543-26914543 on Build GRCh38
rs759391482 Chr.12: 26914516-26914516 on Build GRCh38
rs762104351 Chr.12: 26914488-26914488 on Build GRCh38
rs995715473 Chr.12: 26914652-26914652 on Build GRCh38
rs756851956 Chr.12: 26914574-26914574 on Build GRCh38
rs1015499464 Chr.12: 26914640-26914640 on Build GRCh38
rs899541753 Chr.12: 26914485-26914485 on Build GRCh38
rs903228594 Chr.12: 26914515-26914515 on Build GRCh38
rs144434963 Chr.12: 26914546-26914546 on Build GRCh38
rs148835650 Chr.12: 26914597-26914597 on Build GRCh38
rs780665416 Chr.12: 26914606-26914606 on Build GRCh38
rs768886878 Chr.12: 26914478-26914478 on Build GRCh38

View Full Product Details