Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 105116772-105117021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BVES
Entrez Gene ID
11149
Gene Name
blood vessel epicardial substance
Gene Aliases
HBVES, LGMD2X, POP1, POPDC1
UniGene
Hs.221660
Gene Chromosome Location
Chr.6: 105096822-105137174 on Build GRCh38
Transcripts
NM_001199563.1 NM_007073.4 NM_147147.3 XM_011535398.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6172267 c.677G>T p.R226I Chr.6: 105116840-105116840 on Build GRCh38
4389534 c.744C>A p.I248I Chr.6: 105116773-105116773 on Build GRCh38
3619152 c.701G>A p.R234K Chr.6: 105116816-105116816 on Build GRCh38
1071708 c.703T>C p.L235L Chr.6: 105116814-105116814 on Build GRCh38
6328442 c.697G>T p.E233* Chr.6: 105116820-105116820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61995860 Chr.6: 105116809-105116809 on Build GRCh38
rs371418501 Chr.6: 105116982-105116982 on Build GRCh38
rs982732184 Chr.6: 105116970-105116970 on Build GRCh38
rs758521326 Chr.6: 105116913-105116919 on Build GRCh38
rs1033728924 Chr.6: 105116868-105116868 on Build GRCh38
rs776279592 Chr.6: 105116916-105116916 on Build GRCh38
rs765485974 Chr.6: 105116869-105116871 on Build GRCh38
rs763577934 Chr.6: 105116913-105116913 on Build GRCh38
rs369395137 Chr.6: 105116884-105116884 on Build GRCh38
rs557964721 Chr.6: 105116927-105116927 on Build GRCh38
rs1033835926 Chr.6: 105116992-105116992 on Build GRCh38
rs746333757 Chr.6: 105116826-105116826 on Build GRCh38
rs752769453 Chr.6: 105116890-105116890 on Build GRCh38
rs1024212854 Chr.6: 105116933-105116933 on Build GRCh38
rs774873887 Chr.6: 105116811-105116811 on Build GRCh38
rs944619247 Chr.6: 105116830-105116830 on Build GRCh38
rs765032691 Chr.6: 105116918-105116918 on Build GRCh38
rs749543466 Chr.6: 105116805-105116805 on Build GRCh38
rs763235125 Chr.6: 105116806-105116806 on Build GRCh38
rs145419931 Chr.6: 105116893-105116893 on Build GRCh38
rs772673239 Chr.6: 105116883-105116883 on Build GRCh38
rs971744415 Chr.6: 105116958-105116958 on Build GRCh38
rs991317388 Chr.6: 105116905-105116905 on Build GRCh38
rs113922292 Chr.6: 105116825-105116825 on Build GRCh38

View Full Product Details