Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74028119-74028346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELN
Entrez Gene ID
2006
Gene Name
elastin
Gene Aliases
SVAS, WBS, WS
UniGene
Hs.647061
Gene Chromosome Location
Chr.7: 74027772-74069907 on Build GRCh38
Transcripts
NM_000501.3 NM_001081752.2 NM_001081753.2 NM_001081754.2 NM_001081755.2 NM_001278912.1 NM_001278913.1 NM_001278914.1 NM_001278915.1 NM_001278916.1 NM_001278917.1 NM_001278918.1 NM_001278939.1 XM_005250187.1 XM_005250188.1 XM_011515868.1 XM_011515869.1 XM_011515870.1 XM_011515871.1 XM_011515872.1 XM_011515873.1 XM_011515874.1 XM_011515875.1 XM_011515876.1 XM_011515877.1 XM_017011813.1 XM_017011814.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5873042 c.7G>A p.G3S Chr.7: 74028194-74028194 on Build GRCh38
5407864 c.3G>A p.M1I Chr.7: 74028190-74028190 on Build GRCh38
6530140 c.54G>C p.L18L Chr.7: 74028241-74028241 on Build GRCh38
5873041 c.7G>A p.G3S Chr.7: 74028194-74028194 on Build GRCh38
6653461 c.29G>A p.R10Q Chr.7: 74028216-74028216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782267385 Chr.7: 74028151-74028151 on Build GRCh38
rs782339711 Chr.7: 74028158-74028158 on Build GRCh38
rs373874745 Chr.7: 74028298-74028298 on Build GRCh38
rs782223292 Chr.7: 74028215-74028215 on Build GRCh38
rs781829148 Chr.7: 74028181-74028181 on Build GRCh38
rs782452483 Chr.7: 74028202-74028202 on Build GRCh38
rs370874654 Chr.7: 74028297-74028297 on Build GRCh38
rs782483220 Chr.7: 74028248-74028248 on Build GRCh38
rs781894708 Chr.7: 74028255-74028255 on Build GRCh38
rs782540463 Chr.7: 74028193-74028193 on Build GRCh38
rs782196032 Chr.7: 74028157-74028157 on Build GRCh38
rs782153925 Chr.7: 74028308-74028308 on Build GRCh38
rs41410045 Chr.7: 74028150-74028150 on Build GRCh38
rs781784045 Chr.7: 74028201-74028201 on Build GRCh38
rs782284924 Chr.7: 74028167-74028167 on Build GRCh38
rs727503022 Chr.7: 74028230-74028231 on Build GRCh38
rs782389304 Chr.7: 74028151-74028152 on Build GRCh38
rs782568692 Chr.7: 74028156-74028156 on Build GRCh38
rs782750950 Chr.7: 74028314-74028314 on Build GRCh38
rs781987539 Chr.7: 74028181-74028182 on Build GRCh38
rs559210020 Chr.7: 74028207-74028207 on Build GRCh38
rs931132347 Chr.7: 74028275-74028275 on Build GRCh38
rs782411752 Chr.7: 74028153-74028153 on Build GRCh38
rs782448509 Chr.7: 74028313-74028313 on Build GRCh38
rs781942461 Chr.7: 74028214-74028214 on Build GRCh38
rs782133424 Chr.7: 74028164-74028164 on Build GRCh38
rs782047987 Chr.7: 74028175-74028175 on Build GRCh38
rs782666754 Chr.7: 74028196-74028196 on Build GRCh38
rs559112365 Chr.7: 74028143-74028143 on Build GRCh38
rs781964320 Chr.7: 74028162-74028162 on Build GRCh38
rs782107954 Chr.7: 74028184-74028184 on Build GRCh38
rs782601747 Chr.7: 74028204-74028204 on Build GRCh38
rs782814385 Chr.7: 74028311-74028311 on Build GRCh38
rs781820838 Chr.7: 74028236-74028236 on Build GRCh38
rs142370984 Chr.7: 74028256-74028256 on Build GRCh38
rs781978295 Chr.7: 74028304-74028304 on Build GRCh38
rs782252362 Chr.7: 74028272-74028272 on Build GRCh38
rs782365678 Chr.7: 74028218-74028218 on Build GRCh38
rs782737546 Chr.7: 74028300-74028300 on Build GRCh38
rs149127344 Chr.7: 74028221-74028221 on Build GRCh38
rs373864057 Chr.7: 74028220-74028220 on Build GRCh38
rs782292334 Chr.7: 74028213-74028213 on Build GRCh38
rs45569239 Chr.7: 74028148-74028148 on Build GRCh38
rs781792091 Chr.7: 74028312-74028312 on Build GRCh38
rs569660857 Chr.7: 74028208-74028208 on Build GRCh38
rs782726436 Chr.7: 74028224-74028224 on Build GRCh38
rs541115449 Chr.7: 74028179-74028179 on Build GRCh38
rs782761490 Chr.7: 74028251-74028251 on Build GRCh38
rs781993784 Chr.7: 74028219-74028219 on Build GRCh38
rs542540015 Chr.7: 74028205-74028205 on Build GRCh38
rs782766792 Chr.7: 74028192-74028192 on Build GRCh38
rs863223518 Chr.7: 74028189-74028189 on Build GRCh38
rs151272992 Chr.7: 74028261-74028261 on Build GRCh38

View Full Product Details