Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 80362890-80363147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYDC2
Entrez Gene ID
84332
Gene Name
DPY30 domain containing 2
Gene Aliases
-
UniGene
Hs.741500
Gene Chromosome Location
Chr.10: 80344745-80368073 on Build GRCh38
Transcripts
NM_001270041.1 NM_001270042.1 NM_032372.5 NR_070308.1 NR_070309.1 XM_011540267.2 XM_011540268.1 XM_011540270.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3751999 c.234G>A p.E78E Chr.10: 80363037-80363037 on Build GRCh38
5201000 c.190-1G>C p.? Chr.10: 80362950-80362950 on Build GRCh38
3666784 c.209T>C p.M70T Chr.10: 80363012-80363012 on Build GRCh38
3665913 c.230A>G p.E77G Chr.10: 80363033-80363033 on Build GRCh38
3867941 c.154G>A p.E52K Chr.10: 80362957-80362957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759136576 Chr.10: 80362926-80362926 on Build GRCh38
rs756130606 Chr.10: 80363099-80363099 on Build GRCh38
rs748217257 Chr.10: 80362990-80362990 on Build GRCh38
rs755252460 Chr.10: 80363084-80363084 on Build GRCh38
rs764132041 Chr.10: 80363055-80363055 on Build GRCh38
rs764944790 Chr.10: 80363074-80363074 on Build GRCh38
rs7903783 Chr.10: 80362949-80362949 on Build GRCh38
rs202043386 Chr.10: 80363114-80363114 on Build GRCh38
rs199911588 Chr.10: 80362962-80362962 on Build GRCh38
rs767094128 Chr.10: 80362934-80362934 on Build GRCh38
rs778115670 Chr.10: 80363006-80363006 on Build GRCh38
rs200074369 Chr.10: 80363021-80363021 on Build GRCh38
rs386745754 Chr.10: 80363120-80363124 on Build GRCh38
rs774174247 Chr.10: 80363031-80363031 on Build GRCh38
rs1035147748 Chr.10: 80362945-80362945 on Build GRCh38
rs749715497 Chr.10: 80363008-80363008 on Build GRCh38
rs752817348 Chr.10: 80363098-80363098 on Build GRCh38
rs770401553 Chr.10: 80363000-80363000 on Build GRCh38
rs909613319 Chr.10: 80362993-80362993 on Build GRCh38
rs747303304 Chr.10: 80362952-80362952 on Build GRCh38
rs774127450 Chr.10: 80362924-80362924 on Build GRCh38
rs372569725 Chr.10: 80362931-80362931 on Build GRCh38
rs201475044 Chr.10: 80363124-80363125 on Build GRCh38
rs1000255872 Chr.10: 80362917-80362917 on Build GRCh38
rs559035656 Chr.10: 80362981-80362983 on Build GRCh38
rs771149006 Chr.10: 80363017-80363017 on Build GRCh38
rs544093696 Chr.10: 80363126-80363126 on Build GRCh38
rs965674138 Chr.10: 80363065-80363065 on Build GRCh38
rs567841615 Chr.10: 80363035-80363035 on Build GRCh38
rs375922844 Chr.10: 80362947-80362947 on Build GRCh38
rs753696739 Chr.10: 80363066-80363066 on Build GRCh38
rs575243045 Chr.10: 80363122-80363122 on Build GRCh38
rs763892300 Chr.10: 80363122-80363123 on Build GRCh38
rs149794065 Chr.10: 80363111-80363124 on Build GRCh38
rs751742005 Chr.10: 80362956-80362956 on Build GRCh38
rs558273591 Chr.10: 80363121-80363121 on Build GRCh38
rs763923793 Chr.10: 80362943-80362943 on Build GRCh38
rs778369711 Chr.10: 80363113-80363113 on Build GRCh38
rs538590290 Chr.10: 80363120-80363120 on Build GRCh38
rs751795115 Chr.10: 80363081-80363081 on Build GRCh38
rs759532681 Chr.10: 80362945-80362946 on Build GRCh38
rs757940558 Chr.10: 80363120-80363121 on Build GRCh38
rs1538818 Chr.10: 80363037-80363037 on Build GRCh38
rs933869601 Chr.10: 80363061-80363061 on Build GRCh38
rs553079164 Chr.10: 80363052-80363052 on Build GRCh38
rs761700638 Chr.10: 80363073-80363073 on Build GRCh38
rs777998251 Chr.10: 80362936-80362936 on Build GRCh38
rs752283542 Chr.10: 80363016-80363017 on Build GRCh38
rs3765582 Chr.10: 80362983-80362983 on Build GRCh38
rs112746297 Chr.10: 80362941-80362941 on Build GRCh38
rs781391925 Chr.10: 80363090-80363090 on Build GRCh38
rs770695940 Chr.10: 80362969-80362969 on Build GRCh38
rs766098774 Chr.10: 80362919-80362919 on Build GRCh38
rs760097230 Chr.10: 80363053-80363053 on Build GRCh38

View Full Product Details