Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6935841-6936098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATN1
Entrez Gene ID
1822
Gene Name
atrophin 1
Gene Aliases
B37, D12S755E, DRPLA, HRS, NOD
UniGene
Hs.143766
Gene Chromosome Location
Chr.12: 6924463-6942321 on Build GRCh38
Transcripts
NM_001007026.1 NM_001940.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
267888 c.712G>C p.G238R Chr.12: 6935979-6935979 on Build GRCh38
4044562 c.805G>A p.G269S Chr.12: 6936072-6936072 on Build GRCh38
6606935 c.736G>A p.G246S Chr.12: 6936003-6936003 on Build GRCh38
4502352 c.610C>T p.R204* Chr.12: 6935877-6935877 on Build GRCh38
1705007 c.589C>T p.P197S Chr.12: 6935856-6935856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782018994 Chr.12: 6936037-6936037 on Build GRCh38
rs782639608 Chr.12: 6935896-6935896 on Build GRCh38
rs782696666 Chr.12: 6935982-6935982 on Build GRCh38
rs782295854 Chr.12: 6935902-6935902 on Build GRCh38
rs782028005 Chr.12: 6935935-6935935 on Build GRCh38
rs781803346 Chr.12: 6935872-6935872 on Build GRCh38
rs782490677 Chr.12: 6935881-6935881 on Build GRCh38
rs782237952 Chr.12: 6936004-6936004 on Build GRCh38
rs745377685 Chr.12: 6936012-6936012 on Build GRCh38
rs782509730 Chr.12: 6935890-6935890 on Build GRCh38
rs145331713 Chr.12: 6936005-6936005 on Build GRCh38
rs1017313697 Chr.12: 6936059-6936059 on Build GRCh38
rs781792190 Chr.12: 6935975-6935975 on Build GRCh38
rs782344779 Chr.12: 6935922-6935922 on Build GRCh38
rs782233355 Chr.12: 6935914-6935914 on Build GRCh38
rs1017103579 Chr.12: 6935991-6935991 on Build GRCh38
rs149811742 Chr.12: 6935964-6935964 on Build GRCh38
rs139909258 Chr.12: 6935997-6935997 on Build GRCh38
rs148694613 Chr.12: 6936015-6936015 on Build GRCh38
rs782046159 Chr.12: 6936039-6936039 on Build GRCh38
rs782267292 Chr.12: 6935983-6935983 on Build GRCh38
rs782804407 Chr.12: 6936036-6936036 on Build GRCh38
rs782333025 Chr.12: 6935992-6935992 on Build GRCh38
rs782004113 Chr.12: 6935924-6935924 on Build GRCh38
rs782745379 Chr.12: 6936070-6936070 on Build GRCh38
rs782715706 Chr.12: 6935974-6935974 on Build GRCh38
rs782422818 Chr.12: 6935933-6935933 on Build GRCh38
rs782605849 Chr.12: 6935977-6935977 on Build GRCh38
rs781823465 Chr.12: 6936072-6936072 on Build GRCh38
rs143136221 Chr.12: 6936069-6936069 on Build GRCh38
rs781901082 Chr.12: 6935889-6935889 on Build GRCh38
rs781916976 Chr.12: 6935953-6935953 on Build GRCh38
rs782222553 Chr.12: 6935980-6935980 on Build GRCh38
rs371645176 Chr.12: 6935870-6935870 on Build GRCh38
rs781849378 Chr.12: 6936040-6936040 on Build GRCh38
rs782516322 Chr.12: 6936078-6936078 on Build GRCh38
rs782119229 Chr.12: 6935928-6935928 on Build GRCh38
rs782693165 Chr.12: 6935944-6935944 on Build GRCh38
rs782061811 Chr.12: 6935943-6935943 on Build GRCh38
rs782759433 Chr.12: 6936080-6936080 on Build GRCh38
rs782075942 Chr.12: 6935958-6935958 on Build GRCh38
rs200736794 Chr.12: 6935981-6935981 on Build GRCh38

View Full Product Details