Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 97226307-97226552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA6
Entrez Gene ID
285220
Gene Name
EPH receptor A6
Gene Aliases
EHK-2, EHK2, EK12, EPA6, HEK12, PRO57066
UniGene
Hs.272208
Gene Chromosome Location
Chr.3: 96814581-97761532 on Build GRCh38
Transcripts
NM_001080448.2 XM_006713592.3 XM_017006210.1 XM_017006211.1 XM_017006212.1 XM_017006213.1 XM_017006214.1 XM_017006219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3598051 c.951C>T p.F317F Chr.3: 97226382-97226382 on Build GRCh38
3598048 c.892C>T p.P298S Chr.3: 97226323-97226323 on Build GRCh38
5455236 c.962A>G p.K321R Chr.3: 97226393-97226393 on Build GRCh38
4681311 c.1265G>C p.C422S Chr.3: 97226414-97226414 on Build GRCh38
1425839 c.1179C>T p.H393H Chr.3: 97226328-97226328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758462964 Chr.3: 97226355-97226355 on Build GRCh38
rs185161556 Chr.3: 97226501-97226501 on Build GRCh38
rs777717850 Chr.3: 97226357-97226357 on Build GRCh38
rs375090191 Chr.3: 97226388-97226388 on Build GRCh38
rs778590340 Chr.3: 97226386-97226386 on Build GRCh38
rs995764547 Chr.3: 97226515-97226515 on Build GRCh38
rs752883948 Chr.3: 97226354-97226354 on Build GRCh38
rs771605845 Chr.3: 97226411-97226411 on Build GRCh38
rs369809638 Chr.3: 97226391-97226391 on Build GRCh38
rs538135505 Chr.3: 97226485-97226485 on Build GRCh38
rs756098664 Chr.3: 97226365-97226365 on Build GRCh38
rs747423146 Chr.3: 97226435-97226435 on Build GRCh38
rs779816245 Chr.3: 97226368-97226368 on Build GRCh38
rs768362929 Chr.3: 97226370-97226370 on Build GRCh38
rs765543671 Chr.3: 97226341-97226341 on Build GRCh38
rs1016123144 Chr.3: 97226502-97226502 on Build GRCh38
rs910812697 Chr.3: 97226348-97226348 on Build GRCh38
rs866282511 Chr.3: 97226352-97226352 on Build GRCh38
rs777112210 Chr.3: 97226427-97226427 on Build GRCh38
rs779518542 Chr.3: 97226460-97226460 on Build GRCh38
rs752894799 Chr.3: 97226451-97226451 on Build GRCh38
rs750300622 Chr.3: 97226362-97226362 on Build GRCh38
rs575076714 Chr.3: 97226511-97226511 on Build GRCh38
rs372156304 Chr.3: 97226429-97226429 on Build GRCh38
rs200172139 Chr.3: 97226439-97226439 on Build GRCh38
rs749134413 Chr.3: 97226369-97226369 on Build GRCh38
rs761260100 Chr.3: 97226407-97226407 on Build GRCh38

View Full Product Details