Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 42917374-42917647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPSTI1
Entrez Gene ID
94240
Gene Name
epithelial stromal interaction 1 (breast)
Gene Aliases
BRESI1
UniGene
Hs.546467
Gene Chromosome Location
Chr.13: 42886388-42992271 on Build GRCh38
Transcripts
NM_001002264.2 NM_033255.3 XM_005266596.1 XM_005266597.1 XM_006719896.3 XM_011535312.1 XM_011535313.1 XM_011535314.1 XM_017020853.1 XM_017020854.1 XM_017020855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3676597 c.774+2T>A p.? Chr.13: 42917539-42917539 on Build GRCh38
5432574 c.691-10A>T p.? Chr.13: 42917634-42917634 on Build GRCh38
696541 c.698G>C p.S233T Chr.13: 42917617-42917617 on Build GRCh38
4827332 c.750G>C p.K250N Chr.13: 42917565-42917565 on Build GRCh38
947563 c.730G>T p.E244* Chr.13: 42917585-42917585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747674213 Chr.13: 42917516-42917516 on Build GRCh38
rs761835922 Chr.13: 42917550-42917550 on Build GRCh38
rs1044986566 Chr.13: 42917449-42917449 on Build GRCh38
rs753433367 Chr.13: 42917597-42917597 on Build GRCh38
rs540409181 Chr.13: 42917408-42917408 on Build GRCh38
rs757354005 Chr.13: 42917614-42917615 on Build GRCh38
rs778411217 Chr.13: 42917513-42917513 on Build GRCh38
rs760972340 Chr.13: 42917467-42917467 on Build GRCh38
rs779631527 Chr.13: 42917501-42917501 on Build GRCh38
rs763280095 Chr.13: 42917536-42917536 on Build GRCh38
rs554086177 Chr.13: 42917476-42917476 on Build GRCh38
rs773552657 Chr.13: 42917453-42917453 on Build GRCh38
rs770873975 Chr.13: 42917530-42917530 on Build GRCh38
rs1048372484 Chr.13: 42917609-42917609 on Build GRCh38
rs776871406 Chr.13: 42917533-42917533 on Build GRCh38
rs369471861 Chr.13: 42917607-42917607 on Build GRCh38
rs754360491 Chr.13: 42917548-42917550 on Build GRCh38
rs146505612 Chr.13: 42917564-42917564 on Build GRCh38
rs778607555 Chr.13: 42917604-42917604 on Build GRCh38
rs141768190 Chr.13: 42917601-42917601 on Build GRCh38
rs201357180 Chr.13: 42917540-42917540 on Build GRCh38
rs934406522 Chr.13: 42917472-42917472 on Build GRCh38
rs748967645 Chr.13: 42917511-42917511 on Build GRCh38
rs780405135 Chr.13: 42917572-42917572 on Build GRCh38
rs746952721 Chr.13: 42917619-42917619 on Build GRCh38
rs372074142 Chr.13: 42917508-42917508 on Build GRCh38
rs1010243645 Chr.13: 42917502-42917502 on Build GRCh38
rs546045106 Chr.13: 42917520-42917520 on Build GRCh38
rs747664866 Chr.13: 42917606-42917606 on Build GRCh38
rs562190581 Chr.13: 42917557-42917557 on Build GRCh38
rs375655517 Chr.13: 42917512-42917512 on Build GRCh38
rs756427611 Chr.13: 42917571-42917571 on Build GRCh38
rs531697485 Chr.13: 42917541-42917541 on Build GRCh38
rs761366036 Chr.13: 42917584-42917584 on Build GRCh38
rs777718724 Chr.13: 42917611-42917611 on Build GRCh38
rs760498726 Chr.13: 42917526-42917526 on Build GRCh38
rs541981219 Chr.13: 42917543-42917543 on Build GRCh38
rs76537292 Chr.13: 42917478-42917478 on Build GRCh38

View Full Product Details