Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65581975-65582238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHBP1L1
Entrez Gene ID
254102
Gene Name
EH domain binding protein 1 like 1
Gene Aliases
-
UniGene
Hs.502867
Gene Chromosome Location
Chr.11: 65576034-65592650 on Build GRCh38
Transcripts
NM_001099409.1 XM_005273867.3 XM_005273868.3 XM_005273869.3 XM_005273870.3 XM_006718486.3 XM_006718487.3 XM_017017467.1 XM_017017468.1 XM_017017469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3676266 c.1359G>T p.G453G Chr.11: 65582031-65582031 on Build GRCh38
5983928 c.1528G>A p.E510K Chr.11: 65582200-65582200 on Build GRCh38
3451831 c.1424C>T p.P475L Chr.11: 65582096-65582096 on Build GRCh38
7365989 c.1559G>T p.G520V Chr.11: 65582231-65582231 on Build GRCh38
1355963 c.1513_1514delGA p.E507fs*12 Chr.11: 65582185-65582186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573095833 Chr.11: 65582131-65582131 on Build GRCh38
rs757792722 Chr.11: 65582112-65582112 on Build GRCh38
rs1057505016 Chr.11: 65582062-65582062 on Build GRCh38
rs747575909 Chr.11: 65582011-65582011 on Build GRCh38
rs773127626 Chr.11: 65582053-65582053 on Build GRCh38
rs771371262 Chr.11: 65582017-65582017 on Build GRCh38
rs760698202 Chr.11: 65582057-65582057 on Build GRCh38
rs759516296 Chr.11: 65582158-65582158 on Build GRCh38
rs556715013 Chr.11: 65582096-65582096 on Build GRCh38
rs545610877 Chr.11: 65582100-65582100 on Build GRCh38
rs937948596 Chr.11: 65582142-65582142 on Build GRCh38
rs752362522 Chr.11: 65582095-65582095 on Build GRCh38
rs117879560 Chr.11: 65582031-65582031 on Build GRCh38
rs780517300 Chr.11: 65582147-65582147 on Build GRCh38
rs537597997 Chr.11: 65582070-65582070 on Build GRCh38
rs763202364 Chr.11: 65582051-65582051 on Build GRCh38
rs746365198 Chr.11: 65582121-65582121 on Build GRCh38
rs1015664668 Chr.11: 65582208-65582208 on Build GRCh38
rs753336840 Chr.11: 65581996-65581996 on Build GRCh38
rs778387395 Chr.11: 65581998-65581998 on Build GRCh38
rs576660708 Chr.11: 65582099-65582099 on Build GRCh38
rs754393486 Chr.11: 65582082-65582082 on Build GRCh38
rs76222487 Chr.11: 65582033-65582033 on Build GRCh38
rs552820442 Chr.11: 65582124-65582124 on Build GRCh38
rs370393705 Chr.11: 65581997-65581997 on Build GRCh38
rs764716500 Chr.11: 65582197-65582197 on Build GRCh38
rs368726387 Chr.11: 65582092-65582092 on Build GRCh38
rs7945803 Chr.11: 65582211-65582211 on Build GRCh38
rs776697433 Chr.11: 65582022-65582022 on Build GRCh38
rs754919584 Chr.11: 65582001-65582002 on Build GRCh38
rs753417724 Chr.11: 65582074-65582074 on Build GRCh38
rs781629750 Chr.11: 65582116-65582116 on Build GRCh38
rs752197650 Chr.11: 65582213-65582213 on Build GRCh38
rs749531094 Chr.11: 65582130-65582130 on Build GRCh38
rs541419920 Chr.11: 65582157-65582157 on Build GRCh38
rs778110943 Chr.11: 65582010-65582010 on Build GRCh38
rs190443164 Chr.11: 65581995-65581995 on Build GRCh38
rs758052033 Chr.11: 65582215-65582215 on Build GRCh38
rs767486768 Chr.11: 65582052-65582052 on Build GRCh38
rs745898424 Chr.11: 65582025-65582025 on Build GRCh38
rs747309216 Chr.11: 65582106-65582106 on Build GRCh38
rs748564593 Chr.11: 65582005-65582005 on Build GRCh38
rs373711828 Chr.11: 65582007-65582007 on Build GRCh38
rs775633565 Chr.11: 65582125-65582125 on Build GRCh38
rs779704700 Chr.11: 65582134-65582134 on Build GRCh38
rs372365243 Chr.11: 65582154-65582154 on Build GRCh38
rs778752732 Chr.11: 65582060-65582061 on Build GRCh38

View Full Product Details