Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144080533-144080770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC4
Entrez Gene ID
54512
Gene Name
exosome component 4
Gene Aliases
RRP41, RRP41A, Rrp41p, SKI6, Ski6p, hRrp41p, p12A
UniGene
Hs.632041
Gene Chromosome Location
Chr.8: 144064092-144080648 on Build GRCh38
Transcripts
NM_019037.2 XM_011517134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6657303 c.689G>A p.R230Q Chr.8: 144080552-144080552 on Build GRCh38
5884669 c.732G>A p.G244G Chr.8: 144080595-144080595 on Build GRCh38
749645 c.674A>T p.H225L Chr.8: 144080537-144080537 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782613941 Chr.8: 144080634-144080634 on Build GRCh38
rs782499711 Chr.8: 144080623-144080623 on Build GRCh38
rs564119007 Chr.8: 144080636-144080636 on Build GRCh38
rs782721445 Chr.8: 144080592-144080592 on Build GRCh38
rs949014794 Chr.8: 144080728-144080728 on Build GRCh38
rs182925683 Chr.8: 144080573-144080573 on Build GRCh38
rs982830636 Chr.8: 144080631-144080631 on Build GRCh38
rs938687062 Chr.8: 144080666-144080666 on Build GRCh38
rs367990298 Chr.8: 144080567-144080567 on Build GRCh38
rs765969412 Chr.8: 144080647-144080647 on Build GRCh38
rs781819179 Chr.8: 144080631-144080631 on Build GRCh38
rs781872196 Chr.8: 144080616-144080616 on Build GRCh38
rs782407812 Chr.8: 144080651-144080651 on Build GRCh38
rs140746867 Chr.8: 144080747-144080747 on Build GRCh38
rs894708297 Chr.8: 144080700-144080700 on Build GRCh38
rs150345900 Chr.8: 144080584-144080584 on Build GRCh38
rs782316494 Chr.8: 144080561-144080561 on Build GRCh38
rs782036452 Chr.8: 144080564-144080564 on Build GRCh38
rs782567906 Chr.8: 144080641-144080641 on Build GRCh38
rs113485259 Chr.8: 144080570-144080570 on Build GRCh38
rs782458306 Chr.8: 144080640-144080640 on Build GRCh38
rs1017514720 Chr.8: 144080574-144080574 on Build GRCh38
rs1057451096 Chr.8: 144080691-144080691 on Build GRCh38
rs751112751 Chr.8: 144080662-144080662 on Build GRCh38
rs782805058 Chr.8: 144080612-144080612 on Build GRCh38
rs781789176 Chr.8: 144080594-144080594 on Build GRCh38
rs782220917 Chr.8: 144080560-144080560 on Build GRCh38

View Full Product Details