Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55269262-55269489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX29
Entrez Gene ID
54505
Gene Name
DEAH-box helicase 29
Gene Aliases
DDX29
UniGene
Hs.593268
Gene Chromosome Location
Chr.5: 55256245-55307693 on Build GRCh38
Transcripts
NM_019030.2 XM_006714653.2 XM_017009588.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
40020 c.3252G>A p.K1084K Chr.5: 55269455-55269455 on Build GRCh38
1068815 c.3272T>A p.I1091K Chr.5: 55269435-55269435 on Build GRCh38
5011407 c.3253A>T p.M1085L Chr.5: 55269454-55269454 on Build GRCh38
5988802 c.3266G>T p.G1089V Chr.5: 55269441-55269441 on Build GRCh38
1068817 c.3218T>G p.L1073R Chr.5: 55269489-55269489 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs571237310 Chr.5: 55269402-55269402 on Build GRCh38
rs567552868 Chr.5: 55269382-55269382 on Build GRCh38
rs752640484 Chr.5: 55269368-55269368 on Build GRCh38
rs1054705281 Chr.5: 55269339-55269339 on Build GRCh38
rs953175151 Chr.5: 55269418-55269418 on Build GRCh38
rs954515271 Chr.5: 55269371-55269371 on Build GRCh38
rs965866604 Chr.5: 55269343-55269343 on Build GRCh38
rs1022644882 Chr.5: 55269310-55269310 on Build GRCh38
rs527571369 Chr.5: 55269327-55269327 on Build GRCh38
rs11958488 Chr.5: 55269355-55269355 on Build GRCh38
rs1017432163 Chr.5: 55269456-55269456 on Build GRCh38
rs780597164 Chr.5: 55269416-55269416 on Build GRCh38
rs541418209 Chr.5: 55269309-55269309 on Build GRCh38
rs113714433 Chr.5: 55269298-55269298 on Build GRCh38
rs929287283 Chr.5: 55269286-55269286 on Build GRCh38
rs372698671 Chr.5: 55269422-55269422 on Build GRCh38
rs201861427 Chr.5: 55269389-55269389 on Build GRCh38
rs192022239 Chr.5: 55269315-55269315 on Build GRCh38
rs565202621 Chr.5: 55269285-55269285 on Build GRCh38
rs200658088 Chr.5: 55269396-55269396 on Build GRCh38
rs749108815 Chr.5: 55269340-55269340 on Build GRCh38
rs532675951 Chr.5: 55269299-55269299 on Build GRCh38
rs756329979 Chr.5: 55269405-55269405 on Build GRCh38
rs758196802 Chr.5: 55269378-55269378 on Build GRCh38
rs975853329 Chr.5: 55269352-55269352 on Build GRCh38
rs751373744 Chr.5: 55269388-55269388 on Build GRCh38
rs1011551155 Chr.5: 55269302-55269302 on Build GRCh38
rs999368984 Chr.5: 55269326-55269326 on Build GRCh38
rs749702719 Chr.5: 55269452-55269452 on Build GRCh38

View Full Product Details