Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120655862-120656135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP1
Entrez Gene ID
9478
Gene Name
calcium binding protein 1
Gene Aliases
CALBRAIN, HCALB_BR
UniGene
Hs.458482
Gene Chromosome Location
Chr.12: 120640552-120680869 on Build GRCh38
Transcripts
NM_001033677.1 NM_004276.4 NM_031205.3 XM_017020235.1 XM_017020236.1 XM_017020237.1 XM_017020238.1 XM_017020239.1

SNP Detail

SNP ID Chromosome Location
rs771530368 Chr.12: 120655975-120655975 on Build GRCh38
rs767345122 Chr.12: 120656019-120656019 on Build GRCh38
rs753729072 Chr.12: 120655925-120655925 on Build GRCh38
rs749857611 Chr.12: 120655967-120655967 on Build GRCh38
rs764132379 Chr.12: 120656006-120656006 on Build GRCh38
rs567864309 Chr.12: 120656081-120656081 on Build GRCh38
rs747654115 Chr.12: 120656049-120656049 on Build GRCh38
rs774918900 Chr.12: 120655976-120655976 on Build GRCh38
rs750233148 Chr.12: 120656020-120656020 on Build GRCh38
rs750215119 Chr.12: 120655939-120655939 on Build GRCh38
rs778445492 Chr.12: 120655965-120655965 on Build GRCh38
rs570197529 Chr.12: 120655936-120655936 on Build GRCh38
rs760834876 Chr.12: 120656005-120656005 on Build GRCh38
rs1027812981 Chr.12: 120655892-120655892 on Build GRCh38
rs766582579 Chr.12: 120655974-120655974 on Build GRCh38
rs772279321 Chr.12: 120656078-120656078 on Build GRCh38
rs754635466 Chr.12: 120656048-120656048 on Build GRCh38
rs948710625 Chr.12: 120655984-120655984 on Build GRCh38
rs751122848 Chr.12: 120656038-120656038 on Build GRCh38
rs766135094 Chr.12: 120656112-120656112 on Build GRCh38
rs759802299 Chr.12: 120655977-120655977 on Build GRCh38
rs577446843 Chr.12: 120656095-120656095 on Build GRCh38
rs772347438 Chr.12: 120655994-120655994 on Build GRCh38
rs760608374 Chr.12: 120655917-120655917 on Build GRCh38
rs767606603 Chr.12: 120655896-120655896 on Build GRCh38
rs753886376 Chr.12: 120656012-120656012 on Build GRCh38
rs956330586 Chr.12: 120655906-120655906 on Build GRCh38
rs552282303 Chr.12: 120655956-120655956 on Build GRCh38
rs758061514 Chr.12: 120656034-120656034 on Build GRCh38
rs149552643 Chr.12: 120656094-120656094 on Build GRCh38
rs764126077 Chr.12: 120655919-120655919 on Build GRCh38
rs144243264 Chr.12: 120656108-120656108 on Build GRCh38
rs746279211 Chr.12: 120656077-120656077 on Build GRCh38
rs775538231 Chr.12: 120655998-120655998 on Build GRCh38
rs757206110 Chr.12: 120656020-120656020 on Build GRCh38
rs761788616 Chr.12: 120656011-120656011 on Build GRCh38
rs765164207 Chr.12: 120656105-120656105 on Build GRCh38
rs368608595 Chr.12: 120656080-120656080 on Build GRCh38
rs565809078 Chr.12: 120656054-120656054 on Build GRCh38
rs779514753 Chr.12: 120655943-120655943 on Build GRCh38
rs759908451 Chr.12: 120655891-120655891 on Build GRCh38
rs752839613 Chr.12: 120655907-120655907 on Build GRCh38
rs768009622 Chr.12: 120655960-120655960 on Build GRCh38
rs997830740 Chr.12: 120655922-120655923 on Build GRCh38
rs757167024 Chr.12: 120655934-120655934 on Build GRCh38
rs766086473 Chr.12: 120656035-120656035 on Build GRCh38
rs762828769 Chr.12: 120656110-120656110 on Build GRCh38
rs746532714 Chr.12: 120655955-120655955 on Build GRCh38
rs539208630 Chr.12: 120655941-120655941 on Build GRCh38
rs571435428 Chr.12: 120656050-120656050 on Build GRCh38
rs779499562 Chr.12: 120656062-120656062 on Build GRCh38

View Full Product Details