Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 83198390-83198643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD42
Entrez Gene ID
338699
Gene Name
ankyrin repeat domain 42
Gene Aliases
PPP1R79, SARP
UniGene
Hs.503438
Gene Chromosome Location
Chr.11: 83193739-83256099 on Build GRCh38
Transcripts
NM_001300972.1 NM_001300973.1 NM_001300975.1 NM_001300976.1 NM_001300977.1 NM_182603.3 NR_125355.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4201305 c.87C>T p.S29S Chr.11: 83198507-83198507 on Build GRCh38
4201304 c.59-4A>G p.? Chr.11: 83198475-83198475 on Build GRCh38
5814575 c.92A>G p.H31R Chr.11: 83198512-83198512 on Build GRCh38
5766473 c.103C>T p.R35* Chr.11: 83198523-83198523 on Build GRCh38
5814574 c.92A>G p.H31R Chr.11: 83198512-83198512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777859004 Chr.11: 83198512-83198512 on Build GRCh38
rs764757828 Chr.11: 83198457-83198457 on Build GRCh38
rs770654416 Chr.11: 83198553-83198553 on Build GRCh38
rs779177252 Chr.11: 83198508-83198508 on Build GRCh38
rs187835644 Chr.11: 83198475-83198475 on Build GRCh38
rs754202663 Chr.11: 83198613-83198613 on Build GRCh38
rs777535581 Chr.11: 83198492-83198493 on Build GRCh38
rs190623446 Chr.11: 83198562-83198562 on Build GRCh38
rs376131866 Chr.11: 83198451-83198451 on Build GRCh38
rs370470773 Chr.11: 83198436-83198436 on Build GRCh38
rs144724340 Chr.11: 83198555-83198555 on Build GRCh38
rs750868327 Chr.11: 83198480-83198480 on Build GRCh38
rs200776346 Chr.11: 83198432-83198432 on Build GRCh38
rs754328040 Chr.11: 83198494-83198494 on Build GRCh38
rs770687899 Chr.11: 83198437-83198437 on Build GRCh38
rs761481836 Chr.11: 83198605-83198605 on Build GRCh38
rs567974633 Chr.11: 83198488-83198488 on Build GRCh38
rs201002064 Chr.11: 83198476-83198476 on Build GRCh38
rs535783567 Chr.11: 83198506-83198506 on Build GRCh38
rs745831097 Chr.11: 83198561-83198561 on Build GRCh38
rs775040014 Chr.11: 83198563-83198563 on Build GRCh38
rs766687726 Chr.11: 83198608-83198608 on Build GRCh38
rs760176304 Chr.11: 83198617-83198617 on Build GRCh38
rs776446935 Chr.11: 83198448-83198448 on Build GRCh38
rs1023043241 Chr.11: 83198600-83198600 on Build GRCh38
rs748222729 Chr.11: 83198509-83198509 on Build GRCh38
rs765831741 Chr.11: 83198620-83198620 on Build GRCh38
rs772398117 Chr.11: 83198429-83198429 on Build GRCh38
rs1015512619 Chr.11: 83198543-83198543 on Build GRCh38
rs367717761 Chr.11: 83198517-83198517 on Build GRCh38
rs762446701 Chr.11: 83198584-83198584 on Build GRCh38
rs150340749 Chr.11: 83198422-83198423 on Build GRCh38
rs773893068 Chr.11: 83198596-83198596 on Build GRCh38
rs768087844 Chr.11: 83198592-83198592 on Build GRCh38
rs377381576 Chr.11: 83198523-83198523 on Build GRCh38
rs756439669 Chr.11: 83198482-83198482 on Build GRCh38
rs758757336 Chr.11: 83198510-83198510 on Build GRCh38
rs199536647 Chr.11: 83198574-83198574 on Build GRCh38

View Full Product Details