Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29900992-29901259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPHD1
Entrez Gene ID
253982
Gene Name
aspartate beta-hydroxylase domain containing 1
Gene Aliases
-
UniGene
Hs.655761
Gene Chromosome Location
Chr.16: 29900392-29919860 on Build GRCh38
Transcripts
NM_181718.3 XM_017023107.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1609188 c.24C>T p.F8F Chr.16: 29900995-29900995 on Build GRCh38
7198078 c.173C>A p.P58Q Chr.16: 29901144-29901144 on Build GRCh38
5954235 c.209C>T p.P70L Chr.16: 29901180-29901180 on Build GRCh38
4151304 c.53G>T p.R18I Chr.16: 29901024-29901024 on Build GRCh38
4925661 c.274C>T p.L92F Chr.16: 29901245-29901245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148255395 Chr.16: 29901205-29901205 on Build GRCh38
rs763725447 Chr.16: 29901191-29901191 on Build GRCh38
rs141266563 Chr.16: 29901220-29901220 on Build GRCh38
rs759359640 Chr.16: 29901139-29901139 on Build GRCh38
rs767276871 Chr.16: 29901020-29901023 on Build GRCh38
rs766267066 Chr.16: 29901017-29901017 on Build GRCh38
rs760072559 Chr.16: 29901180-29901180 on Build GRCh38
rs748760695 Chr.16: 29901209-29901209 on Build GRCh38
rs1002297583 Chr.16: 29901085-29901085 on Build GRCh38
rs369015409 Chr.16: 29901029-29901030 on Build GRCh38
rs747388392 Chr.16: 29901076-29901076 on Build GRCh38
rs775183731 Chr.16: 29901223-29901224 on Build GRCh38
rs777851583 Chr.16: 29901223-29901223 on Build GRCh38
rs923456071 Chr.16: 29901201-29901201 on Build GRCh38
rs373933675 Chr.16: 29901159-29901159 on Build GRCh38
rs762628760 Chr.16: 29901012-29901012 on Build GRCh38
rs146783719 Chr.16: 29901099-29901099 on Build GRCh38
rs751485896 Chr.16: 29901020-29901020 on Build GRCh38
rs773306612 Chr.16: 29901121-29901121 on Build GRCh38
rs767392159 Chr.16: 29901149-29901149 on Build GRCh38
rs148270044 Chr.16: 29901193-29901193 on Build GRCh38
rs753568043 Chr.16: 29901050-29901050 on Build GRCh38
rs781771679 Chr.16: 29901089-29901089 on Build GRCh38
rs754852635 Chr.16: 29901024-29901024 on Build GRCh38
rs376294577 Chr.16: 29901129-29901129 on Build GRCh38
rs771145770 Chr.16: 29901232-29901232 on Build GRCh38
rs780521617 Chr.16: 29901070-29901070 on Build GRCh38
rs570295575 Chr.16: 29901136-29901136 on Build GRCh38
rs376342563 Chr.16: 29901128-29901128 on Build GRCh38
rs751928998 Chr.16: 29901198-29901198 on Build GRCh38
rs755671354 Chr.16: 29901043-29901043 on Build GRCh38
rs752678469 Chr.16: 29901179-29901179 on Build GRCh38
rs756871445 Chr.16: 29901197-29901197 on Build GRCh38
rs758928203 Chr.16: 29901153-29901153 on Build GRCh38
rs372770612 Chr.16: 29901042-29901042 on Build GRCh38
rs749333781 Chr.16: 29901227-29901227 on Build GRCh38
rs758684908 Chr.16: 29901054-29901054 on Build GRCh38
rs765278930 Chr.16: 29901080-29901080 on Build GRCh38
rs767449446 Chr.16: 29901032-29901032 on Build GRCh38
rs777266963 Chr.16: 29901047-29901047 on Build GRCh38
rs140635603 Chr.16: 29901111-29901111 on Build GRCh38

View Full Product Details