Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48327450-48327693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMP3
Entrez Gene ID
2014
Gene Name
epithelial membrane protein 3
Gene Aliases
YMP
UniGene
Hs.9999
Gene Chromosome Location
Chr.19: 48325372-48330555 on Build GRCh38
Transcripts
NM_001313905.1 NM_001425.2 XM_011526605.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4563971 c.99G>A p.G33G Chr.19: 48327541-48327541 on Build GRCh38
6317644 c.97G>T p.G33W Chr.19: 48327539-48327539 on Build GRCh38
3721628 c.124G>A p.D42N Chr.19: 48327566-48327566 on Build GRCh38
6653887 c.142G>A p.D48N Chr.19: 48327584-48327584 on Build GRCh38
998933 c.141C>T p.N47N Chr.19: 48327583-48327583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751684161 Chr.19: 48327524-48327524 on Build GRCh38
rs75558953 Chr.19: 48327492-48327492 on Build GRCh38
rs535061522 Chr.19: 48327507-48327507 on Build GRCh38
rs919053368 Chr.19: 48327603-48327603 on Build GRCh38
rs1053592752 Chr.19: 48327666-48327666 on Build GRCh38
rs748935769 Chr.19: 48327526-48327527 on Build GRCh38
rs771518677 Chr.19: 48327663-48327663 on Build GRCh38
rs566153158 Chr.19: 48327503-48327503 on Build GRCh38
rs867833586 Chr.19: 48327628-48327628 on Build GRCh38
rs149302468 Chr.19: 48327610-48327610 on Build GRCh38
rs893248888 Chr.19: 48327672-48327672 on Build GRCh38
rs371695295 Chr.19: 48327640-48327640 on Build GRCh38
rs748467464 Chr.19: 48327545-48327545 on Build GRCh38
rs757823214 Chr.19: 48327642-48327642 on Build GRCh38
rs771714548 Chr.19: 48327582-48327582 on Build GRCh38
rs767665615 Chr.19: 48327638-48327638 on Build GRCh38
rs755308681 Chr.19: 48327528-48327528 on Build GRCh38
rs554878809 Chr.19: 48327573-48327573 on Build GRCh38
rs146091933 Chr.19: 48327540-48327540 on Build GRCh38
rs752444529 Chr.19: 48327499-48327499 on Build GRCh38
rs985771577 Chr.19: 48327508-48327508 on Build GRCh38
rs761908174 Chr.19: 48327612-48327612 on Build GRCh38
rs755042868 Chr.19: 48327637-48327637 on Build GRCh38
rs749652705 Chr.19: 48327641-48327641 on Build GRCh38
rs768347751 Chr.19: 48327531-48327532 on Build GRCh38
rs760407687 Chr.19: 48327589-48327589 on Build GRCh38
rs765251929 Chr.19: 48327617-48327617 on Build GRCh38
rs370992861 Chr.19: 48327566-48327566 on Build GRCh38
rs953286217 Chr.19: 48327506-48327506 on Build GRCh38
rs139549534 Chr.19: 48327574-48327574 on Build GRCh38
rs201392469 Chr.19: 48327565-48327565 on Build GRCh38
rs374673796 Chr.19: 48327649-48327649 on Build GRCh38
rs767157000 Chr.19: 48327493-48327493 on Build GRCh38
rs752917182 Chr.19: 48327639-48327639 on Build GRCh38
rs539162973 Chr.19: 48327471-48327471 on Build GRCh38
rs775165167 Chr.19: 48327584-48327584 on Build GRCh38
rs748118595 Chr.19: 48327629-48327641 on Build GRCh38
rs374754875 Chr.19: 48327514-48327514 on Build GRCh38
rs776257972 Chr.19: 48327595-48327595 on Build GRCh38
rs745410009 Chr.19: 48327656-48327656 on Build GRCh38
rs772096719 Chr.19: 48327660-48327661 on Build GRCh38
rs370758994 Chr.19: 48327502-48327502 on Build GRCh38
rs773831068 Chr.19: 48327474-48327474 on Build GRCh38
rs778509172 Chr.19: 48327614-48327614 on Build GRCh38

View Full Product Details