Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 55351208-55351453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFRAL
Entrez Gene ID
389400
Gene Name
GDNF family receptor alpha like
Gene Aliases
C6orf144, GRAL, UNQ9356, bA360D14.1
UniGene
Hs.526967
Gene Chromosome Location
Chr.6: 55327469-55402493 on Build GRCh38
Transcripts
NM_207410.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4566597 c.420_421GG>AA p.D141N Chr.6: 55351302-55351303 on Build GRCh38
3628738 c.390C>T p.S130S Chr.6: 55351272-55351272 on Build GRCh38
3628739 c.419G>A p.G140E Chr.6: 55351301-55351301 on Build GRCh38
3628736 c.379G>A p.G127R Chr.6: 55351261-55351261 on Build GRCh38
376866 c.375C>A p.F125L Chr.6: 55351257-55351257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774631165 Chr.6: 55351397-55351397 on Build GRCh38
rs770299137 Chr.6: 55351268-55351268 on Build GRCh38
rs936760204 Chr.6: 55351259-55351259 on Build GRCh38
rs768305198 Chr.6: 55351414-55351414 on Build GRCh38
rs758247040 Chr.6: 55351241-55351241 on Build GRCh38
rs748448067 Chr.6: 55351377-55351377 on Build GRCh38
rs779024147 Chr.6: 55351233-55351233 on Build GRCh38
rs1006424846 Chr.6: 55351415-55351415 on Build GRCh38
rs781515663 Chr.6: 55351234-55351234 on Build GRCh38
rs368896330 Chr.6: 55351253-55351253 on Build GRCh38
rs764836576 Chr.6: 55351426-55351426 on Build GRCh38
rs776126884 Chr.6: 55351422-55351422 on Build GRCh38
rs750354645 Chr.6: 55351327-55351327 on Build GRCh38
rs866995388 Chr.6: 55351369-55351369 on Build GRCh38
rs267601084 Chr.6: 55351301-55351301 on Build GRCh38
rs375909531 Chr.6: 55351333-55351333 on Build GRCh38
rs761569699 Chr.6: 55351424-55351424 on Build GRCh38
rs79683091 Chr.6: 55351371-55351371 on Build GRCh38
rs557504977 Chr.6: 55351367-55351367 on Build GRCh38
rs369163760 Chr.6: 55351403-55351403 on Build GRCh38
rs761253451 Chr.6: 55351322-55351322 on Build GRCh38
rs781304511 Chr.6: 55351374-55351374 on Build GRCh38
rs755216350 Chr.6: 55351258-55351258 on Build GRCh38
rs764891737 Chr.6: 55351326-55351326 on Build GRCh38
rs914090384 Chr.6: 55351286-55351286 on Build GRCh38
rs766266730 Chr.6: 55351331-55351331 on Build GRCh38
rs931622163 Chr.6: 55351364-55351364 on Build GRCh38
rs781280070 Chr.6: 55351265-55351265 on Build GRCh38
rs538104435 Chr.6: 55351290-55351290 on Build GRCh38
rs1008776704 Chr.6: 55351263-55351263 on Build GRCh38
rs776165168 Chr.6: 55351321-55351321 on Build GRCh38
rs751329340 Chr.6: 55351362-55351362 on Build GRCh38
rs756153654 Chr.6: 55351379-55351379 on Build GRCh38
rs755089337 Chr.6: 55351294-55351295 on Build GRCh38
rs371493722 Chr.6: 55351297-55351297 on Build GRCh38
rs749829894 Chr.6: 55351277-55351277 on Build GRCh38
rs142171356 Chr.6: 55351396-55351396 on Build GRCh38
rs777891851 Chr.6: 55351388-55351388 on Build GRCh38
rs774753930 Chr.6: 55351293-55351293 on Build GRCh38
rs749777799 Chr.6: 55351393-55351393 on Build GRCh38
rs185849431 Chr.6: 55351238-55351238 on Build GRCh38
rs773683856 Chr.6: 55351270-55351270 on Build GRCh38
rs758478185 Chr.6: 55351328-55351328 on Build GRCh38
rs200048676 Chr.6: 55351254-55351254 on Build GRCh38

View Full Product Details