Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26448205-26448450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
XM_011514268.2 NM_001242803.1 NM_006994.4 NM_197974.2

Gene Symbol
BTN3A3
Entrez Gene ID
10384
Gene Name
butyrophilin subfamily 3 member A3
Gene Aliases
BTF3, BTN3.3
UniGene
Hs.167741
Gene Chromosome Location
Chr.6: 26440472-26453415 on Build GRCh38
Transcripts
XM_011514268.2 NM_001242803.1 NM_006994.4 NM_197974.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7151423 c.762G>C p.G254G Chr.6: 26448294-26448294 on Build GRCh38
4666402 c.790G>A p.A264T Chr.6: 26448322-26448322 on Build GRCh38
4645562 c.916G>T p.E306* Chr.6: 26448448-26448448 on Build GRCh38
4958573 c.849G>A p.E283E Chr.6: 26448381-26448381 on Build GRCh38
740845 c.863G>T p.R288L Chr.6: 26448395-26448395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756194206 Chr.6: 26448247-26448247 on Build GRCh38
rs780149207 Chr.6: 26448249-26448249 on Build GRCh38
rs371725478 Chr.6: 26448321-26448321 on Build GRCh38
rs375083429 Chr.6: 26448417-26448417 on Build GRCh38
rs760817724 Chr.6: 26448353-26448353 on Build GRCh38
rs759562414 Chr.6: 26448374-26448374 on Build GRCh38
rs764783175 Chr.6: 26448224-26448224 on Build GRCh38
rs867868768 Chr.6: 26448313-26448313 on Build GRCh38
rs990543562 Chr.6: 26448411-26448411 on Build GRCh38
rs140949337 Chr.6: 26448395-26448395 on Build GRCh38
rs972613746 Chr.6: 26448227-26448227 on Build GRCh38
rs371579409 Chr.6: 26448281-26448281 on Build GRCh38
rs200302181 Chr.6: 26448407-26448407 on Build GRCh38
rs200631543 Chr.6: 26448412-26448412 on Build GRCh38
rs748782578 Chr.6: 26448258-26448258 on Build GRCh38
rs41266843 Chr.6: 26448250-26448250 on Build GRCh38
rs764039554 Chr.6: 26448231-26448231 on Build GRCh38
rs750648131 Chr.6: 26448282-26448282 on Build GRCh38
rs868539887 Chr.6: 26448401-26448401 on Build GRCh38
rs763001189 Chr.6: 26448273-26448273 on Build GRCh38
rs144686528 Chr.6: 26448418-26448418 on Build GRCh38
rs368908067 Chr.6: 26448309-26448309 on Build GRCh38
rs779808480 Chr.6: 26448425-26448425 on Build GRCh38
rs753948316 Chr.6: 26448367-26448367 on Build GRCh38
rs770646729 Chr.6: 26448260-26448260 on Build GRCh38
rs1015493065 Chr.6: 26448317-26448317 on Build GRCh38
rs753855926 Chr.6: 26448297-26448297 on Build GRCh38
rs267600913 Chr.6: 26448339-26448339 on Build GRCh38
rs142420060 Chr.6: 26448264-26448264 on Build GRCh38
rs573665384 Chr.6: 26448308-26448308 on Build GRCh38
rs761776152 Chr.6: 26448344-26448344 on Build GRCh38
rs751346138 Chr.6: 26448240-26448240 on Build GRCh38
rs775127231 Chr.6: 26448270-26448270 on Build GRCh38
rs764059839 Chr.6: 26448275-26448275 on Build GRCh38
rs199557026 Chr.6: 26448243-26448243 on Build GRCh38
rs775610269 Chr.6: 26448228-26448228 on Build GRCh38
rs768739996 Chr.6: 26448343-26448343 on Build GRCh38
rs543130389 Chr.6: 26448322-26448322 on Build GRCh38
rs767597946 Chr.6: 26448244-26448244 on Build GRCh38
rs748770253 Chr.6: 26448324-26448324 on Build GRCh38
rs553170648 Chr.6: 26448349-26448349 on Build GRCh38
rs572239197 Chr.6: 26448364-26448364 on Build GRCh38
rs779352287 Chr.6: 26448306-26448306 on Build GRCh38
rs368341416 Chr.6: 26448267-26448267 on Build GRCh38
rs762980566 Chr.6: 26448230-26448230 on Build GRCh38
rs748444480 Chr.6: 26448259-26448259 on Build GRCh38
rs146453476 Chr.6: 26448347-26448347 on Build GRCh38
rs745567370 Chr.6: 26448320-26448320 on Build GRCh38
rs758931047 Chr.6: 26448394-26448394 on Build GRCh38
rs761470254 Chr.6: 26448280-26448280 on Build GRCh38
rs774323832 Chr.6: 26448279-26448279 on Build GRCh38
rs765278522 Chr.6: 26448384-26448384 on Build GRCh38
rs779262268 Chr.6: 26448252-26448252 on Build GRCh38
rs755479102 Chr.6: 26448302-26448302 on Build GRCh38
rs753212790 Chr.6: 26448388-26448388 on Build GRCh38
rs151329283 Chr.6: 26448265-26448265 on Build GRCh38
rs760945316 Chr.6: 26448294-26448294 on Build GRCh38

View Full Product Details