Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77428271-77428544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VIPAS39
Entrez Gene ID
63894
Gene Name
VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog
Gene Aliases
C14orf133, SPE-39, SPE39, VIPAR, VPS16B, hSPE-39
UniGene
Hs.16157
Gene Chromosome Location
Chr.14: 77426675-77457640 on Build GRCh38
Transcripts
NM_001193314.1 NM_001193315.1 NM_001193316.1 NM_001193317.1 NM_022067.3 XM_011537066.1 XM_017021579.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3706404 c.1395A>G p.A465A Chr.14: 77428436-77428436 on Build GRCh38
2139403 c.1379G>A p.R460H Chr.14: 77428452-77428452 on Build GRCh38
4437007 c.1411G>C p.D471H Chr.14: 77428420-77428420 on Build GRCh38
3498242 c.1378C>T p.R460C Chr.14: 77428453-77428453 on Build GRCh38
7397307 c.1437G>C p.K479N Chr.14: 77428394-77428394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200474619 Chr.14: 77428460-77428460 on Build GRCh38
rs766947055 Chr.14: 77428358-77428358 on Build GRCh38
rs35622669 Chr.14: 77428292-77428293 on Build GRCh38
rs768536529 Chr.14: 77428506-77428506 on Build GRCh38
rs751596992 Chr.14: 77428327-77428328 on Build GRCh38
rs905180434 Chr.14: 77428386-77428386 on Build GRCh38
rs376541848 Chr.14: 77428382-77428382 on Build GRCh38
rs901072810 Chr.14: 77428304-77428304 on Build GRCh38
rs749965150 Chr.14: 77428482-77428482 on Build GRCh38
rs756669400 Chr.14: 77428357-77428357 on Build GRCh38
rs754289597 Chr.14: 77428360-77428360 on Build GRCh38
rs773189629 Chr.14: 77428435-77428435 on Build GRCh38
rs779979806 Chr.14: 77428495-77428495 on Build GRCh38
rs186485228 Chr.14: 77428515-77428515 on Build GRCh38
rs767491725 Chr.14: 77428334-77428334 on Build GRCh38
rs762011296 Chr.14: 77428325-77428325 on Build GRCh38
rs755447534 Chr.14: 77428457-77428457 on Build GRCh38
rs766112659 Chr.14: 77428453-77428453 on Build GRCh38
rs947875205 Chr.14: 77428342-77428342 on Build GRCh38
rs138274958 Chr.14: 77428438-77428438 on Build GRCh38
rs368874079 Chr.14: 77428497-77428497 on Build GRCh38
rs755554295 Chr.14: 77428493-77428493 on Build GRCh38
rs749085304 Chr.14: 77428392-77428392 on Build GRCh38
rs755331201 Chr.14: 77428370-77428370 on Build GRCh38
rs780846411 Chr.14: 77428472-77428472 on Build GRCh38
rs769744188 Chr.14: 77428384-77428384 on Build GRCh38
rs1029102328 Chr.14: 77428514-77428514 on Build GRCh38
rs558959240 Chr.14: 77428431-77428431 on Build GRCh38
rs376479671 Chr.14: 77428407-77428407 on Build GRCh38
rs766517989 Chr.14: 77428323-77428326 on Build GRCh38
rs140365206 Chr.14: 77428371-77428371 on Build GRCh38
rs892057602 Chr.14: 77428387-77428387 on Build GRCh38
rs113067724 Chr.14: 77428509-77428509 on Build GRCh38
rs768277423 Chr.14: 77428394-77428394 on Build GRCh38
rs75211061 Chr.14: 77428381-77428381 on Build GRCh38
rs866847808 Chr.14: 77428397-77428399 on Build GRCh38
rs756855024 Chr.14: 77428379-77428379 on Build GRCh38
rs201273507 Chr.14: 77428467-77428467 on Build GRCh38
rs750980625 Chr.14: 77428348-77428348 on Build GRCh38
rs775365343 Chr.14: 77428388-77428388 on Build GRCh38
rs754342872 Chr.14: 77428452-77428452 on Build GRCh38
rs765399312 Chr.14: 77428339-77428339 on Build GRCh38
rs150958425 Chr.14: 77428351-77428351 on Build GRCh38
rs759346738 Chr.14: 77428451-77428451 on Build GRCh38
rs145453157 Chr.14: 77428376-77428376 on Build GRCh38
rs773504364 Chr.14: 77428522-77428522 on Build GRCh38
rs149168488 Chr.14: 77428468-77428468 on Build GRCh38

View Full Product Details