Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 67076493-67076720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSPP1
Entrez Gene ID
79848
Gene Name
centrosome and spindle pole associated protein 1
Gene Aliases
CSPP, JBTS21
UniGene
Hs.370147
Gene Chromosome Location
Chr.8: 67062417-67196614 on Build GRCh38
Transcripts
NM_024790.6 XM_005251305.3 XM_006716474.2 XM_006716477.2 XM_011517598.1 XM_011517599.1 XM_011517600.1 XM_011517601.1 XM_011517602.1 XM_011517603.1 XM_011517607.1 XM_011517608.1 XM_017013847.1 XM_017013848.1 XM_017013849.1 XM_017013850.1 XM_017013851.1 XM_017013852.1 XM_017013853.1 XM_017013854.1 XM_017013855.1 XM_017013856.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1190804 c.265_266delGA p.E89fs*6 Chr.8: 67076539-67076540 on Build GRCh38
3834933 c.222G>A p.A74A Chr.8: 67076496-67076496 on Build GRCh38
5812883 c.219A>T p.S73S Chr.8: 67076493-67076493 on Build GRCh38
6226280 c.249G>T p.L83L Chr.8: 67076523-67076523 on Build GRCh38
3901127 c.225G>A p.K75K Chr.8: 67076499-67076499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs180685142 Chr.8: 67076565-67076565 on Build GRCh38
rs765095778 Chr.8: 67076548-67076548 on Build GRCh38
rs747835628 Chr.8: 67076682-67076682 on Build GRCh38
rs143887656 Chr.8: 67076655-67076655 on Build GRCh38
rs771751921 Chr.8: 67076629-67076629 on Build GRCh38
rs760570129 Chr.8: 67076520-67076520 on Build GRCh38
rs773073443 Chr.8: 67076629-67076630 on Build GRCh38
rs757503066 Chr.8: 67076633-67076633 on Build GRCh38
rs369327242 Chr.8: 67076624-67076624 on Build GRCh38
rs1031564673 Chr.8: 67076622-67076622 on Build GRCh38
rs188323303 Chr.8: 67076549-67076549 on Build GRCh38
rs865842077 Chr.8: 67076521-67076521 on Build GRCh38
rs999113119 Chr.8: 67076619-67076619 on Build GRCh38
rs767674652 Chr.8: 67076569-67076569 on Build GRCh38
rs997479629 Chr.8: 67076649-67076649 on Build GRCh38
rs375985758 Chr.8: 67076530-67076530 on Build GRCh38
rs779468454 Chr.8: 67076622-67076623 on Build GRCh38
rs773848283 Chr.8: 67076625-67076626 on Build GRCh38
rs747905557 Chr.8: 67076626-67076627 on Build GRCh38
rs776584930 Chr.8: 67076529-67076529 on Build GRCh38
rs748822329 Chr.8: 67076624-67076625 on Build GRCh38
rs752873157 Chr.8: 67076570-67076570 on Build GRCh38
rs756283251 Chr.8: 67076594-67076594 on Build GRCh38
rs768536004 Chr.8: 67076526-67076526 on Build GRCh38
rs1012663082 Chr.8: 67076544-67076544 on Build GRCh38

View Full Product Details