Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 3850146-3850391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM50B
Entrez Gene ID
26240
Gene Name
family with sequence similarity 50 member B
Gene Aliases
D6S2654E, X5L
UniGene
Hs.140944
Gene Chromosome Location
Chr.6: 3831901-3851320 on Build GRCh38
Transcripts
NM_012135.2 XM_017010729.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6659811 c.472C>T p.R158C Chr.6: 3850283-3850283 on Build GRCh38
3873636 c.547G>A p.E183K Chr.6: 3850358-3850358 on Build GRCh38
1718816 c.481G>A p.E161K Chr.6: 3850292-3850292 on Build GRCh38
3873635 c.395A>G p.D132G Chr.6: 3850206-3850206 on Build GRCh38
6659810 c.382G>A p.D128N Chr.6: 3850193-3850193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776304566 Chr.6: 3850221-3850221 on Build GRCh38
rs760539577 Chr.6: 3850217-3850217 on Build GRCh38
rs764611763 Chr.6: 3850315-3850315 on Build GRCh38
rs780894143 Chr.6: 3850357-3850357 on Build GRCh38
rs764464170 Chr.6: 3850184-3850184 on Build GRCh38
rs751666178 Chr.6: 3850184-3850186 on Build GRCh38
rs756536862 Chr.6: 3850240-3850240 on Build GRCh38
rs765571239 Chr.6: 3850317-3850317 on Build GRCh38
rs548182725 Chr.6: 3850308-3850308 on Build GRCh38
rs758218604 Chr.6: 3850270-3850270 on Build GRCh38
rs143961511 Chr.6: 3850205-3850205 on Build GRCh38
rs768080374 Chr.6: 3850226-3850226 on Build GRCh38
rs937947723 Chr.6: 3850212-3850212 on Build GRCh38
rs761289588 Chr.6: 3850222-3850222 on Build GRCh38
rs1038694972 Chr.6: 3850324-3850324 on Build GRCh38
rs982004752 Chr.6: 3850192-3850192 on Build GRCh38
rs371839744 Chr.6: 3850326-3850326 on Build GRCh38
rs1005809067 Chr.6: 3850369-3850369 on Build GRCh38
rs372461028 Chr.6: 3850327-3850327 on Build GRCh38
rs757488297 Chr.6: 3850316-3850316 on Build GRCh38
rs373712054 Chr.6: 3850223-3850223 on Build GRCh38
rs1045548785 Chr.6: 3850256-3850256 on Build GRCh38
rs752106061 Chr.6: 3850175-3850175 on Build GRCh38
rs781474180 Chr.6: 3850180-3850180 on Build GRCh38
rs201214003 Chr.6: 3850249-3850249 on Build GRCh38
rs1049964016 Chr.6: 3850343-3850343 on Build GRCh38
rs371366147 Chr.6: 3850194-3850194 on Build GRCh38
rs746031683 Chr.6: 3850281-3850282 on Build GRCh38
rs745851693 Chr.6: 3850214-3850214 on Build GRCh38
rs762548695 Chr.6: 3850300-3850300 on Build GRCh38
rs754166929 Chr.6: 3850246-3850246 on Build GRCh38
rs747860750 Chr.6: 3850285-3850285 on Build GRCh38
rs892917112 Chr.6: 3850304-3850304 on Build GRCh38
rs750428859 Chr.6: 3850319-3850319 on Build GRCh38
rs536693606 Chr.6: 3850279-3850279 on Build GRCh38
rs766840357 Chr.6: 3850165-3850165 on Build GRCh38
rs776107993 Chr.6: 3850312-3850312 on Build GRCh38
rs772952711 Chr.6: 3850366-3850366 on Build GRCh38
rs997731871 Chr.6: 3850338-3850338 on Build GRCh38
rs769437441 Chr.6: 3850286-3850286 on Build GRCh38
rs989854826 Chr.6: 3850228-3850228 on Build GRCh38
rs769385982 Chr.6: 3850365-3850365 on Build GRCh38
rs200167230 Chr.6: 3850314-3850314 on Build GRCh38
rs370896067 Chr.6: 3850278-3850278 on Build GRCh38
rs770121174 Chr.6: 3850286-3850288 on Build GRCh38
rs926532304 Chr.6: 3850207-3850207 on Build GRCh38
rs754758966 Chr.6: 3850355-3850355 on Build GRCh38
rs553770846 Chr.6: 3850348-3850348 on Build GRCh38
rs745966134 Chr.6: 3850267-3850267 on Build GRCh38
rs778167500 Chr.6: 3850204-3850204 on Build GRCh38
rs780070639 Chr.6: 3850273-3850273 on Build GRCh38
rs755352309 Chr.6: 3850177-3850177 on Build GRCh38
rs775081957 Chr.6: 3850216-3850216 on Build GRCh38
rs759890689 Chr.6: 3850224-3850224 on Build GRCh38
rs34635612 Chr.6: 3850229-3850229 on Build GRCh38
rs768152468 Chr.6: 3850218-3850218 on Build GRCh38
rs757273644 Chr.6: 3850256-3850257 on Build GRCh38
rs771178963 Chr.6: 3850210-3850210 on Build GRCh38
rs778114421 Chr.6: 3850245-3850245 on Build GRCh38
rs778972917 Chr.6: 3850213-3850213 on Build GRCh38
rs781281732 Chr.6: 3850274-3850279 on Build GRCh38
rs767874546 Chr.6: 3850306-3850306 on Build GRCh38
rs756235222 Chr.6: 3850199-3850199 on Build GRCh38
rs915800900 Chr.6: 3850234-3850234 on Build GRCh38
rs747947650 Chr.6: 3850358-3850358 on Build GRCh38
rs746748541 Chr.6: 3850277-3850277 on Build GRCh38
rs772536143 Chr.6: 3850298-3850298 on Build GRCh38

View Full Product Details