Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 78241631-78241868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP78
Entrez Gene ID
84131
Gene Name
centrosomal protein 78
Gene Aliases
C9orf81, IP63
UniGene
Hs.374421
Gene Chromosome Location
Chr.9: 78236062-78278431 on Build GRCh38
Transcripts
NM_001098802.1 NM_032171.1 XM_005252263.4 XM_005252265.2 XM_005252266.4 XM_017015192.1 XM_017015193.1 XM_017015194.1 XM_017015195.1 XM_017015196.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4164044 c.529A>T p.T177S Chr.9: 78241725-78241725 on Build GRCh38
4164041 c.526A>C p.I176L Chr.9: 78241722-78241722 on Build GRCh38
3996618 c.512G>T p.G171V Chr.9: 78241708-78241708 on Build GRCh38
5228096 c.563T>C p.L188P Chr.9: 78241759-78241759 on Build GRCh38
1497129 c.500-2A>T p.? Chr.9: 78241694-78241694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763843219 Chr.9: 78241791-78241791 on Build GRCh38
rs1057517695 Chr.9: 78241730-78241730 on Build GRCh38
rs886686367 Chr.9: 78241813-78241813 on Build GRCh38
rs1052404112 Chr.9: 78241810-78241810 on Build GRCh38
rs914542434 Chr.9: 78241684-78241684 on Build GRCh38
rs779354620 Chr.9: 78241710-78241710 on Build GRCh38
rs775163853 Chr.9: 78241779-78241779 on Build GRCh38
rs767178604 Chr.9: 78241670-78241670 on Build GRCh38
rs745944134 Chr.9: 78241718-78241718 on Build GRCh38
rs373409352 Chr.9: 78241816-78241816 on Build GRCh38
rs199823711 Chr.9: 78241677-78241678 on Build GRCh38
rs758972075 Chr.9: 78241832-78241832 on Build GRCh38
rs78373498 Chr.9: 78241686-78241686 on Build GRCh38
rs755863103 Chr.9: 78241677-78241677 on Build GRCh38
rs368674833 Chr.9: 78241732-78241732 on Build GRCh38
rs766897249 Chr.9: 78241778-78241778 on Build GRCh38
rs760345902 Chr.9: 78241785-78241785 on Build GRCh38
rs774205882 Chr.9: 78241766-78241766 on Build GRCh38
rs751079527 Chr.9: 78241661-78241661 on Build GRCh38
rs977625054 Chr.9: 78241700-78241700 on Build GRCh38
rs757369088 Chr.9: 78241678-78241678 on Build GRCh38
rs948743121 Chr.9: 78241833-78241833 on Build GRCh38
rs780055186 Chr.9: 78241838-78241838 on Build GRCh38
rs900596669 Chr.9: 78241730-78241730 on Build GRCh38
rs759057765 Chr.9: 78241768-78241768 on Build GRCh38
rs370478499 Chr.9: 78241735-78241735 on Build GRCh38
rs577004260 Chr.9: 78241678-78241679 on Build GRCh38
rs200560864 Chr.9: 78241825-78241825 on Build GRCh38
rs757714365 Chr.9: 78241708-78241708 on Build GRCh38
rs752437934 Chr.9: 78241676-78241676 on Build GRCh38
rs759027765 Chr.9: 78241662-78241662 on Build GRCh38
rs778233243 Chr.9: 78241687-78241687 on Build GRCh38
rs757617837 Chr.9: 78241820-78241820 on Build GRCh38
rs772928824 Chr.9: 78241741-78241741 on Build GRCh38
rs749150559 Chr.9: 78241752-78241752 on Build GRCh38
rs770891882 Chr.9: 78241758-78241758 on Build GRCh38
rs776660214 Chr.9: 78241775-78241775 on Build GRCh38
rs765750353 Chr.9: 78241823-78241823 on Build GRCh38
rs780213877 Chr.9: 78241722-78241722 on Build GRCh38
rs754267465 Chr.9: 78241698-78241698 on Build GRCh38
rs377654767 Chr.9: 78241723-78241723 on Build GRCh38
rs373045691 Chr.9: 78241678-78241678 on Build GRCh38
rs758454353 Chr.9: 78241720-78241720 on Build GRCh38

View Full Product Details