Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196669710-196669965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC150
Entrez Gene ID
284992
Gene Name
coiled-coil domain containing 150
Gene Aliases
-
UniGene
Hs.132519
Gene Chromosome Location
Chr.2: 196639632-196733668 on Build GRCh38
Transcripts
NM_001080539.1 XM_006712438.3 XM_011510985.2 XM_011510987.2 XM_011510988.2 XM_017003861.1 XM_017003862.1 XM_017003863.1 XM_017003864.1 XM_017003865.1 XM_017003866.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5578247 c.914C>T p.S305F Chr.2: 196669854-196669854 on Build GRCh38
337096 c.895T>C p.S299P Chr.2: 196669835-196669835 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs992159355 Chr.2: 196669765-196669767 on Build GRCh38
rs528584263 Chr.2: 196669931-196669931 on Build GRCh38
rs749253835 Chr.2: 196669882-196669883 on Build GRCh38
rs761702753 Chr.2: 196669890-196669890 on Build GRCh38
rs773212580 Chr.2: 196669908-196669908 on Build GRCh38
rs1045513582 Chr.2: 196669893-196669893 on Build GRCh38
rs766384076 Chr.2: 196669829-196669829 on Build GRCh38
rs529366143 Chr.2: 196669770-196669770 on Build GRCh38
rs760745760 Chr.2: 196669827-196669827 on Build GRCh38
rs778971956 Chr.2: 196669838-196669838 on Build GRCh38
rs758639895 Chr.2: 196669843-196669843 on Build GRCh38
rs948151698 Chr.2: 196669811-196669811 on Build GRCh38
rs750284212 Chr.2: 196669813-196669813 on Build GRCh38
rs998753483 Chr.2: 196669924-196669924 on Build GRCh38
rs761492977 Chr.2: 196669806-196669806 on Build GRCh38
rs753152370 Chr.2: 196669840-196669840 on Build GRCh38
rs903012652 Chr.2: 196669899-196669899 on Build GRCh38
rs917878691 Chr.2: 196669797-196669797 on Build GRCh38
rs763837424 Chr.2: 196669791-196669791 on Build GRCh38
rs768476484 Chr.2: 196669863-196669863 on Build GRCh38
rs771694793 Chr.2: 196669902-196669902 on Build GRCh38
rs903917780 Chr.2: 196669831-196669831 on Build GRCh38
rs372180744 Chr.2: 196669836-196669836 on Build GRCh38
rs376829483 Chr.2: 196669917-196669917 on Build GRCh38
rs546961108 Chr.2: 196669744-196669744 on Build GRCh38
rs761554605 Chr.2: 196669854-196669854 on Build GRCh38
rs201570929 Chr.2: 196669830-196669830 on Build GRCh38
rs771552640 Chr.2: 196669850-196669850 on Build GRCh38
rs759859809 Chr.2: 196669773-196669774 on Build GRCh38
rs760659791 Chr.2: 196669910-196669910 on Build GRCh38
rs778349332 Chr.2: 196669846-196669846 on Build GRCh38
rs773828026 Chr.2: 196669871-196669871 on Build GRCh38
rs538059664 Chr.2: 196669928-196669928 on Build GRCh38
rs747418854 Chr.2: 196669847-196669847 on Build GRCh38
rs753958533 Chr.2: 196669919-196669919 on Build GRCh38
rs193222574 Chr.2: 196669754-196669754 on Build GRCh38
rs1032889914 Chr.2: 196669933-196669933 on Build GRCh38
rs767515900 Chr.2: 196669808-196669808 on Build GRCh38
rs774245980 Chr.2: 196669800-196669800 on Build GRCh38
rs150406735 Chr.2: 196669857-196669857 on Build GRCh38

View Full Product Details