Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123325918-123326147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD2
Entrez Gene ID
29028
Gene Name
ATPase family, AAA domain containing 2
Gene Aliases
ANCCA, CT137, PRO2000
UniGene
Hs.370834
Gene Chromosome Location
Chr.8: 123319850-123396992 on Build GRCh38
Transcripts
NM_014109.3 XM_011516994.2 XM_011516996.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3166304 c.3909G>T p.Q1303H Chr.8: 123325986-123325986 on Build GRCh38
1183857 c.3931A>G p.T1311A Chr.8: 123325964-123325964 on Build GRCh38
3166305 c.3906C>T p.S1302S Chr.8: 123325989-123325989 on Build GRCh38
6606362 c.3950C>T p.A1317V Chr.8: 123325945-123325945 on Build GRCh38
366707 c.3937G>C p.E1313Q Chr.8: 123325958-123325958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771987539 Chr.8: 123325994-123325994 on Build GRCh38
rs981305705 Chr.8: 123326037-123326037 on Build GRCh38
rs751897630 Chr.8: 123325940-123325940 on Build GRCh38
rs180764801 Chr.8: 123326113-123326113 on Build GRCh38
rs189878754 Chr.8: 123326040-123326040 on Build GRCh38
rs770621563 Chr.8: 123326015-123326015 on Build GRCh38
rs915222654 Chr.8: 123326107-123326107 on Build GRCh38
rs761603093 Chr.8: 123326093-123326093 on Build GRCh38
rs367692163 Chr.8: 123325998-123325998 on Build GRCh38
rs747679854 Chr.8: 123325990-123325990 on Build GRCh38
rs931536918 Chr.8: 123326066-123326066 on Build GRCh38
rs951812541 Chr.8: 123325987-123325987 on Build GRCh38
rs778350239 Chr.8: 123325981-123325981 on Build GRCh38
rs765587560 Chr.8: 123326034-123326034 on Build GRCh38
rs776619211 Chr.8: 123326020-123326020 on Build GRCh38
rs768334874 Chr.8: 123325980-123325980 on Build GRCh38
rs946733489 Chr.8: 123326109-123326109 on Build GRCh38
rs60391664 Chr.8: 123326027-123326027 on Build GRCh38
rs990518004 Chr.8: 123325965-123325965 on Build GRCh38
rs757570651 Chr.8: 123325964-123325964 on Build GRCh38
rs200204254 Chr.8: 123326070-123326070 on Build GRCh38
rs759802668 Chr.8: 123326025-123326025 on Build GRCh38
rs762960191 Chr.8: 123326045-123326045 on Build GRCh38
rs776516615 Chr.8: 123326087-123326087 on Build GRCh38
rs371959622 Chr.8: 123326011-123326011 on Build GRCh38
rs774648655 Chr.8: 123326071-123326073 on Build GRCh38
rs1048667906 Chr.8: 123326069-123326069 on Build GRCh38
rs79188010 Chr.8: 123326062-123326062 on Build GRCh38
rs769053683 Chr.8: 123326054-123326054 on Build GRCh38
rs781670659 Chr.8: 123325971-123325971 on Build GRCh38
rs993711425 Chr.8: 123325968-123325968 on Build GRCh38
rs1040730078 Chr.8: 123326112-123326112 on Build GRCh38
rs747032192 Chr.8: 123326002-123326002 on Build GRCh38
rs752007280 Chr.8: 123326078-123326078 on Build GRCh38
rs748914423 Chr.8: 123325975-123325975 on Build GRCh38

View Full Product Details