Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 128758253-128758482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLI1
Entrez Gene ID
2313
Gene Name
Fli-1 proto-oncogene, ETS transcription factor
Gene Aliases
EWSR2, SIC-1
UniGene
Hs.504281
Gene Chromosome Location
Chr.11: 128685263-128813267 on Build GRCh38
Transcripts
NM_001167681.2 NM_001271010.1 NM_001271012.1 NM_002017.4 XM_011542701.2 XM_011542702.1 XM_017017405.1 XM_017017406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6531354 c.173T>C p.I58T Chr.11: 128758269-128758269 on Build GRCh38
94322 c.202C>T p.R68W Chr.11: 128758298-128758298 on Build GRCh38
6867151 c.201G>A p.K67K Chr.11: 128758297-128758297 on Build GRCh38
2150704 c.195C>T p.N65N Chr.11: 128758291-128758291 on Build GRCh38
7266680 c.157C>T p.P53S Chr.11: 128758253-128758253 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186120614 Chr.11: 128758291-128758291 on Build GRCh38
rs4265602 Chr.11: 128758348-128758348 on Build GRCh38
rs983251687 Chr.11: 128758397-128758397 on Build GRCh38
rs574563701 Chr.11: 128758454-128758454 on Build GRCh38
rs764399072 Chr.11: 128758295-128758295 on Build GRCh38
rs1049343223 Chr.11: 128758439-128758439 on Build GRCh38
rs752532580 Chr.11: 128758372-128758372 on Build GRCh38
rs930518725 Chr.11: 128758388-128758388 on Build GRCh38
rs140111384 Chr.11: 128758447-128758447 on Build GRCh38
rs780787816 Chr.11: 128758358-128758358 on Build GRCh38
rs750968869 Chr.11: 128758304-128758304 on Build GRCh38
rs779628662 Chr.11: 128758335-128758335 on Build GRCh38
rs949989530 Chr.11: 128758441-128758441 on Build GRCh38
rs752656842 Chr.11: 128758286-128758286 on Build GRCh38
rs907767378 Chr.11: 128758429-128758429 on Build GRCh38
rs758762735 Chr.11: 128758328-128758328 on Build GRCh38
rs965141057 Chr.11: 128758362-128758362 on Build GRCh38
rs1132722 Chr.11: 128758302-128758302 on Build GRCh38
rs747982607 Chr.11: 128758371-128758371 on Build GRCh38
rs201118331 Chr.11: 128758299-128758299 on Build GRCh38
rs899532384 Chr.11: 128758428-128758428 on Build GRCh38
rs543908687 Chr.11: 128758352-128758352 on Build GRCh38
rs754322278 Chr.11: 128758298-128758298 on Build GRCh38
rs979405206 Chr.11: 128758278-128758278 on Build GRCh38
rs754687449 Chr.11: 128758356-128758356 on Build GRCh38

View Full Product Details