Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66591740-66591999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC87
Entrez Gene ID
55231
Gene Name
coiled-coil domain containing 87
Gene Aliases
-
UniGene
Hs.121072
Gene Chromosome Location
Chr.11: 66590169-66593083 on Build GRCh38
Transcripts
NM_018219.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6312428 c.1229C>A p.A410D Chr.11: 66591787-66591787 on Build GRCh38
4908225 c.1060G>A p.A354T Chr.11: 66591956-66591956 on Build GRCh38
4670059 c.1215C>T p.L405L Chr.11: 66591801-66591801 on Build GRCh38
1200068 c.1109G>A p.R370H Chr.11: 66591907-66591907 on Build GRCh38
3452134 c.1144C>T p.L382L Chr.11: 66591872-66591872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927132168 Chr.11: 66591766-66591766 on Build GRCh38
rs771720516 Chr.11: 66591968-66591968 on Build GRCh38
rs141833535 Chr.11: 66591764-66591764 on Build GRCh38
rs762173012 Chr.11: 66591766-66591767 on Build GRCh38
rs137881283 Chr.11: 66591838-66591838 on Build GRCh38
rs778452891 Chr.11: 66591966-66591966 on Build GRCh38
rs765018973 Chr.11: 66591897-66591897 on Build GRCh38
rs768262949 Chr.11: 66591840-66591840 on Build GRCh38
rs747797538 Chr.11: 66591967-66591967 on Build GRCh38
rs768477307 Chr.11: 66591964-66591964 on Build GRCh38
rs1038303870 Chr.11: 66591819-66591819 on Build GRCh38
rs879082571 Chr.11: 66591771-66591771 on Build GRCh38
rs201117269 Chr.11: 66591777-66591777 on Build GRCh38
rs777766804 Chr.11: 66591907-66591907 on Build GRCh38
rs758747499 Chr.11: 66591855-66591855 on Build GRCh38
rs111997006 Chr.11: 66591908-66591908 on Build GRCh38
rs779190045 Chr.11: 66591785-66591785 on Build GRCh38
rs375465158 Chr.11: 66591828-66591828 on Build GRCh38
rs374330821 Chr.11: 66591848-66591848 on Build GRCh38
rs766979684 Chr.11: 66591880-66591880 on Build GRCh38
rs887199177 Chr.11: 66591921-66591921 on Build GRCh38
rs773886806 Chr.11: 66591874-66591874 on Build GRCh38
rs373320692 Chr.11: 66591963-66591963 on Build GRCh38
rs767088192 Chr.11: 66591856-66591856 on Build GRCh38
rs1056976742 Chr.11: 66591816-66591816 on Build GRCh38
rs142496256 Chr.11: 66591927-66591927 on Build GRCh38
rs1051221128 Chr.11: 66591768-66591781 on Build GRCh38
rs898490727 Chr.11: 66591857-66591857 on Build GRCh38
rs747524308 Chr.11: 66591960-66591960 on Build GRCh38
rs752911174 Chr.11: 66591900-66591900 on Build GRCh38
rs778179008 Chr.11: 66591926-66591926 on Build GRCh38
rs761206869 Chr.11: 66591875-66591875 on Build GRCh38
rs199906600 Chr.11: 66591835-66591835 on Build GRCh38
rs748214053 Chr.11: 66591799-66591799 on Build GRCh38
rs755335832 Chr.11: 66591893-66591893 on Build GRCh38
rs1008372278 Chr.11: 66591962-66591962 on Build GRCh38
rs150538178 Chr.11: 66591975-66591975 on Build GRCh38
rs780039959 Chr.11: 66591959-66591959 on Build GRCh38
rs374929199 Chr.11: 66591836-66591836 on Build GRCh38
rs80219769 Chr.11: 66591831-66591831 on Build GRCh38
rs532353345 Chr.11: 66591891-66591891 on Build GRCh38
rs369805771 Chr.11: 66591905-66591905 on Build GRCh38
rs761280214 Chr.11: 66591851-66591851 on Build GRCh38

View Full Product Details