Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104988882-104989139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf79
Entrez Gene ID
122616
Gene Name
chromosome 14 open reading frame 79
Gene Aliases
-
UniGene
Hs.27183
Gene Chromosome Location
Chr.14: 104985801-105000310 on Build GRCh38
Transcripts
NM_174891.3 XM_005267317.3 XM_005267318.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3369906 c.463C>T p.Q155* Chr.14: 104988982-104988982 on Build GRCh38
3386439 c.561T>A p.H187Q Chr.14: 104989080-104989080 on Build GRCh38
953938 c.441G>T p.E147D Chr.14: 104988960-104988960 on Build GRCh38
953939 c.547G>A p.V183I Chr.14: 104989066-104989066 on Build GRCh38
6209334 c.559C>A p.H187N Chr.14: 104989078-104989078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367618108 Chr.14: 104989047-104989047 on Build GRCh38
rs373570912 Chr.14: 104988969-104988969 on Build GRCh38
rs757807040 Chr.14: 104988974-104988974 on Build GRCh38
rs370531724 Chr.14: 104989012-104989012 on Build GRCh38
rs761439232 Chr.14: 104988999-104988999 on Build GRCh38
rs781742201 Chr.14: 104988987-104988987 on Build GRCh38
rs771455716 Chr.14: 104989006-104989006 on Build GRCh38
rs1028875694 Chr.14: 104989111-104989111 on Build GRCh38
rs749707649 Chr.14: 104989055-104989055 on Build GRCh38
rs767444317 Chr.14: 104988949-104988949 on Build GRCh38
rs770202291 Chr.14: 104988990-104988990 on Build GRCh38
rs772777895 Chr.14: 104989091-104989091 on Build GRCh38
rs757425689 Chr.14: 104989037-104989037 on Build GRCh38
rs763609167 Chr.14: 104988933-104988933 on Build GRCh38
rs775190993 Chr.14: 104988913-104988913 on Build GRCh38
rs764424610 Chr.14: 104989025-104989025 on Build GRCh38
rs376812571 Chr.14: 104989102-104989102 on Build GRCh38
rs760722389 Chr.14: 104989093-104989093 on Build GRCh38
rs751699862 Chr.14: 104989030-104989030 on Build GRCh38
rs201634836 Chr.14: 104989072-104989072 on Build GRCh38
rs766260193 Chr.14: 104988964-104988964 on Build GRCh38
rs753160074 Chr.14: 104988922-104988926 on Build GRCh38
rs757262516 Chr.14: 104988983-104988983 on Build GRCh38
rs376913748 Chr.14: 104988994-104988994 on Build GRCh38
rs775776284 Chr.14: 104988993-104988993 on Build GRCh38
rs1013786532 Chr.14: 104989090-104989090 on Build GRCh38
rs923054599 Chr.14: 104989044-104989044 on Build GRCh38
rs548694506 Chr.14: 104988971-104988971 on Build GRCh38
rs754606419 Chr.14: 104988916-104988916 on Build GRCh38
rs778109212 Chr.14: 104989021-104989028 on Build GRCh38
rs527728804 Chr.14: 104989065-104989065 on Build GRCh38
rs770941596 Chr.14: 104989097-104989097 on Build GRCh38
rs373737894 Chr.14: 104989014-104989014 on Build GRCh38
rs531787654 Chr.14: 104988943-104988943 on Build GRCh38
rs756036802 Chr.14: 104988961-104988961 on Build GRCh38
rs747738995 Chr.14: 104989079-104989079 on Build GRCh38
rs749608156 Chr.14: 104989073-104989073 on Build GRCh38
rs562028663 Chr.14: 104989020-104989020 on Build GRCh38
rs773336623 Chr.14: 104989011-104989011 on Build GRCh38
rs375087463 Chr.14: 104989077-104989077 on Build GRCh38
rs902209346 Chr.14: 104989022-104989022 on Build GRCh38
rs767072021 Chr.14: 104988922-104988929 on Build GRCh38
rs746371532 Chr.14: 104988988-104988988 on Build GRCh38
rs376237464 Chr.14: 104988912-104988912 on Build GRCh38
rs554510970 Chr.14: 104988919-104988919 on Build GRCh38
rs563068420 Chr.14: 104988939-104988939 on Build GRCh38
rs370709711 Chr.14: 104988951-104988951 on Build GRCh38
rs377016992 Chr.14: 104988928-104988932 on Build GRCh38
rs781119607 Chr.14: 104989051-104989051 on Build GRCh38
rs753644884 Chr.14: 104989015-104989015 on Build GRCh38
rs372314065 Chr.14: 104989081-104989081 on Build GRCh38
rs764544101 Chr.14: 104988935-104988935 on Build GRCh38
rs368382350 Chr.14: 104989078-104989078 on Build GRCh38
rs145688822 Chr.14: 104989066-104989066 on Build GRCh38
rs773758438 Chr.14: 104988996-104988996 on Build GRCh38
rs984599029 Chr.14: 104988941-104988941 on Build GRCh38
rs761689231 Chr.14: 104988946-104988946 on Build GRCh38

View Full Product Details